Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150885513-150885764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTHFD1L
Entrez Gene ID
25902
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Gene Aliases
FTHFSDC1, MTC1THFS, dJ292B18.2
UniGene
Hs.591343
Gene Chromosome Location
Chr.6: 150865679-151101887 on Build GRCh38
Transcripts
NM_001242767.1 NM_001242768.1 NM_001242769.1 NM_015440.4 XM_005266907.4 XM_005266910.4 XM_005266911.4 XM_011535729.2 XM_011535730.2 XM_011535731.2 XM_011535732.2 XM_011535733.2 XM_011535734.2 XM_011535737.2 XM_011535738.2 XM_017010702.1 XM_017010703.1 XM_017010704.1 XM_017010705.1 XM_017010706.1 XM_017010707.1 XM_017010708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948166 c.553A>C p.I185L Chr.6: 150885644-150885644 on Build GRCh38
5612113 c.548C>T p.T183I Chr.6: 150885639-150885639 on Build GRCh38
483546 c.594T>C p.C198C Chr.6: 150885685-150885685 on Build GRCh38
3019154 c.616G>A p.A206T Chr.6: 150885707-150885707 on Build GRCh38
4350627 c.582T>A p.D194E Chr.6: 150885673-150885673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761328391 Chr.6: 150885584-150885584 on Build GRCh38
rs904006061 Chr.6: 150885575-150885575 on Build GRCh38
rs1025585852 Chr.6: 150885574-150885574 on Build GRCh38
rs781051380 Chr.6: 150885742-150885742 on Build GRCh38
rs750978722 Chr.6: 150885666-150885666 on Build GRCh38
rs774908616 Chr.6: 150885658-150885658 on Build GRCh38
rs1003534020 Chr.6: 150885577-150885577 on Build GRCh38
rs202128387 Chr.6: 150885710-150885710 on Build GRCh38
rs777464746 Chr.6: 150885603-150885603 on Build GRCh38
rs746872338 Chr.6: 150885733-150885733 on Build GRCh38
rs763116264 Chr.6: 150885644-150885644 on Build GRCh38
rs745419265 Chr.6: 150885559-150885559 on Build GRCh38
rs764491999 Chr.6: 150885655-150885655 on Build GRCh38
rs771856814 Chr.6: 150885594-150885594 on Build GRCh38
rs1001147618 Chr.6: 150885576-150885576 on Build GRCh38
rs767075195 Chr.6: 150885610-150885611 on Build GRCh38
rs912276547 Chr.6: 150885553-150885553 on Build GRCh38
rs764681771 Chr.6: 150885701-150885701 on Build GRCh38
rs768208656 Chr.6: 150885664-150885664 on Build GRCh38
rs745816763 Chr.6: 150885744-150885744 on Build GRCh38
rs762398953 Chr.6: 150885659-150885659 on Build GRCh38
rs750087340 Chr.6: 150885653-150885660 on Build GRCh38
rs114282883 Chr.6: 150885564-150885564 on Build GRCh38
rs758034959 Chr.6: 150885719-150885719 on Build GRCh38
rs775733841 Chr.6: 150885638-150885638 on Build GRCh38
rs867357696 Chr.6: 150885573-150885573 on Build GRCh38
rs753343470 Chr.6: 150885545-150885545 on Build GRCh38
rs373791530 Chr.6: 150885616-150885616 on Build GRCh38
rs757048563 Chr.6: 150885738-150885738 on Build GRCh38
rs777529589 Chr.6: 150885726-150885726 on Build GRCh38
rs754458525 Chr.6: 150885697-150885697 on Build GRCh38
rs759543773 Chr.6: 150885618-150885618 on Build GRCh38
rs377728033 Chr.6: 150885627-150885627 on Build GRCh38
rs769305405 Chr.6: 150885569-150885569 on Build GRCh38
rs746925926 Chr.6: 150885605-150885605 on Build GRCh38

View Full Product Details