Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5151403-5151668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52A1
Entrez Gene ID
23538
Gene Name
olfactory receptor family 52 subfamily A member 1
Gene Aliases
HPFH1OR
UniGene
Hs.553520
Gene Chromosome Location
Chr.11: 5151431-5152369 on Build GRCh38
Transcripts
NM_012375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354972 c.702G>A p.K234K Chr.11: 5151668-5151668 on Build GRCh38
4428484 c.814T>G p.S272A Chr.11: 5151556-5151556 on Build GRCh38
5446375 c.801_802insG p.S269fs*13 Chr.11: 5151568-5151569 on Build GRCh38
429136 c.832C>A p.L278I Chr.11: 5151538-5151538 on Build GRCh38
1604662 c.937delT p.*313fs? Chr.11: 5151433-5151433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751457814 Chr.11: 5151631-5151631 on Build GRCh38
rs770087789 Chr.11: 5151455-5151455 on Build GRCh38
rs866627196 Chr.11: 5151507-5151507 on Build GRCh38
rs10768611 Chr.11: 5151556-5151556 on Build GRCh38
rs765713254 Chr.11: 5151604-5151604 on Build GRCh38
rs892773671 Chr.11: 5151548-5151548 on Build GRCh38
rs112884296 Chr.11: 5151461-5151461 on Build GRCh38
rs746950927 Chr.11: 5151537-5151537 on Build GRCh38
rs749910479 Chr.11: 5151589-5151591 on Build GRCh38
rs758172522 Chr.11: 5151617-5151617 on Build GRCh38
rs763715929 Chr.11: 5151471-5151471 on Build GRCh38
rs955859132 Chr.11: 5151564-5151565 on Build GRCh38
rs773485510 Chr.11: 5151572-5151572 on Build GRCh38
rs758175975 Chr.11: 5151511-5151511 on Build GRCh38
rs962723754 Chr.11: 5151458-5151458 on Build GRCh38
rs750227831 Chr.11: 5151502-5151502 on Build GRCh38
rs753480016 Chr.11: 5151486-5151486 on Build GRCh38
rs781176329 Chr.11: 5151641-5151641 on Build GRCh38
rs866373416 Chr.11: 5151430-5151430 on Build GRCh38
rs933417268 Chr.11: 5151599-5151601 on Build GRCh38
rs781264428 Chr.11: 5151549-5151549 on Build GRCh38
rs879018481 Chr.11: 5151546-5151546 on Build GRCh38
rs979083122 Chr.11: 5151538-5151538 on Build GRCh38
rs754916079 Chr.11: 5151640-5151640 on Build GRCh38
rs371802271 Chr.11: 5151462-5151462 on Build GRCh38
rs748305589 Chr.11: 5151550-5151550 on Build GRCh38
rs112098990 Chr.11: 5151565-5151566 on Build GRCh38
rs546776986 Chr.11: 5151540-5151540 on Build GRCh38
rs369074064 Chr.11: 5151624-5151624 on Build GRCh38
rs1004812337 Chr.11: 5151569-5151569 on Build GRCh38
rs780009374 Chr.11: 5151514-5151514 on Build GRCh38
rs774868743 Chr.11: 5151576-5151576 on Build GRCh38
rs199876523 Chr.11: 5151598-5151598 on Build GRCh38
rs199986389 Chr.11: 5151463-5151463 on Build GRCh38
rs138473302 Chr.11: 5151577-5151577 on Build GRCh38
rs879429856 Chr.11: 5151500-5151500 on Build GRCh38
rs948556302 Chr.11: 5151498-5151498 on Build GRCh38
rs201832620 Chr.11: 5151599-5151599 on Build GRCh38
rs1045602038 Chr.11: 5151534-5151534 on Build GRCh38
rs760188234 Chr.11: 5151466-5151466 on Build GRCh38
rs756971811 Chr.11: 5151489-5151489 on Build GRCh38
rs867500065 Chr.11: 5151590-5151590 on Build GRCh38
rs755039211 Chr.11: 5151542-5151542 on Build GRCh38
rs147176940 Chr.11: 5151575-5151575 on Build GRCh38
rs549766506 Chr.11: 5151513-5151533 on Build GRCh38
rs959796156 Chr.11: 5151441-5151441 on Build GRCh38
rs763628174 Chr.11: 5151585-5151585 on Build GRCh38
rs763926084 Chr.11: 5151445-5151446 on Build GRCh38
rs766997566 Chr.11: 5151581-5151583 on Build GRCh38
rs765047426 Chr.11: 5151497-5151497 on Build GRCh38
rs761465169 Chr.11: 5151601-5151601 on Build GRCh38
rs969960392 Chr.11: 5151529-5151529 on Build GRCh38
rs568222482 Chr.11: 5151541-5151541 on Build GRCh38

View Full Product Details