Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215586314-215586585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
UniGene
Hs.335139
Gene Chromosome Location
Chr.1: 215567185-215621821 on Build GRCh38
Transcripts
NM_001319294.1 NM_001319295.1 NM_016121.4 XM_005273158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053091 c.683C>T p.T228I Chr.1: 215586551-215586551 on Build GRCh38
6061308 c.663G>C p.T221T Chr.1: 215586531-215586531 on Build GRCh38
903810 c.702A>G p.L234L Chr.1: 215586570-215586570 on Build GRCh38
4622515 c.704A>G p.N235S Chr.1: 215586572-215586572 on Build GRCh38
5448411 c.696A>G p.V232V Chr.1: 215586564-215586564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998944288 Chr.1: 215586436-215586436 on Build GRCh38
rs567702709 Chr.1: 215586342-215586342 on Build GRCh38
rs368702528 Chr.1: 215586474-215586474 on Build GRCh38
rs374128463 Chr.1: 215586472-215586472 on Build GRCh38
rs996203811 Chr.1: 215586426-215586426 on Build GRCh38
rs765551961 Chr.1: 215586473-215586473 on Build GRCh38
rs913446060 Chr.1: 215586370-215586370 on Build GRCh38
rs1043203444 Chr.1: 215586387-215586387 on Build GRCh38
rs768093736 Chr.1: 215586453-215586453 on Build GRCh38
rs755901570 Chr.1: 215586546-215586546 on Build GRCh38
rs771749430 Chr.1: 215586460-215586460 on Build GRCh38
rs757525261 Chr.1: 215586501-215586501 on Build GRCh38
rs546948902 Chr.1: 215586432-215586432 on Build GRCh38
rs900618392 Chr.1: 215586395-215586407 on Build GRCh38
rs1002990070 Chr.1: 215586555-215586555 on Build GRCh38
rs567736005 Chr.1: 215586388-215586388 on Build GRCh38
rs901965603 Chr.1: 215586409-215586409 on Build GRCh38
rs745899183 Chr.1: 215586379-215586379 on Build GRCh38
rs372677593 Chr.1: 215586530-215586530 on Build GRCh38
rs773904451 Chr.1: 215586455-215586455 on Build GRCh38
rs370200562 Chr.1: 215586466-215586466 on Build GRCh38
rs779723592 Chr.1: 215586550-215586550 on Build GRCh38
rs943394521 Chr.1: 215586404-215586404 on Build GRCh38
rs764258287 Chr.1: 215586483-215586483 on Build GRCh38
rs751757662 Chr.1: 215586496-215586496 on Build GRCh38
rs367959474 Chr.1: 215586417-215586417 on Build GRCh38
rs375751467 Chr.1: 215586531-215586531 on Build GRCh38
rs759893852 Chr.1: 215586467-215586467 on Build GRCh38
rs536379583 Chr.1: 215586407-215586407 on Build GRCh38
rs763569339 Chr.1: 215586480-215586480 on Build GRCh38
rs947588959 Chr.1: 215586386-215586387 on Build GRCh38
rs749205838 Chr.1: 215586556-215586556 on Build GRCh38
rs566722663 Chr.1: 215586442-215586442 on Build GRCh38
rs893324134 Chr.1: 215586443-215586443 on Build GRCh38
rs200168547 Chr.1: 215586493-215586493 on Build GRCh38
rs550801292 Chr.1: 215586365-215586365 on Build GRCh38
rs761448206 Chr.1: 215586457-215586457 on Build GRCh38

View Full Product Details