Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93612824-93613079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6C
Entrez Gene ID
5146
Gene Name
phosphodiesterase 6C
Gene Aliases
ACHM5, COD4, PDEA2
UniGene
Hs.658121
Gene Chromosome Location
Chr.10: 93612588-93665673 on Build GRCh38
Transcripts
NM_006204.3 XM_017016322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392480 c.197T>C p.L66P Chr.10: 93612922-93612922 on Build GRCh38
4713260 c.205G>A p.V69M Chr.10: 93612930-93612930 on Build GRCh38
7180710 c.221G>A p.G74D Chr.10: 93612946-93612946 on Build GRCh38
198784 c.353delC p.T120fs*23 Chr.10: 93613078-93613078 on Build GRCh38
5953023 c.304C>T p.R102W Chr.10: 93613029-93613029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535099102 Chr.10: 93612886-93612886 on Build GRCh38
rs143356640 Chr.10: 93613009-93613009 on Build GRCh38
rs772318774 Chr.10: 93612983-93612983 on Build GRCh38
rs768209111 Chr.10: 93613028-93613028 on Build GRCh38
rs568930062 Chr.10: 93612930-93612930 on Build GRCh38
rs372972245 Chr.10: 93613024-93613024 on Build GRCh38
rs748280050 Chr.10: 93612980-93612980 on Build GRCh38
rs376967510 Chr.10: 93612992-93612992 on Build GRCh38
rs548437074 Chr.10: 93613036-93613036 on Build GRCh38
rs757473047 Chr.10: 93613047-93613047 on Build GRCh38
rs769506319 Chr.10: 93613035-93613035 on Build GRCh38
rs779065446 Chr.10: 93613048-93613048 on Build GRCh38
rs140469169 Chr.10: 93612944-93612944 on Build GRCh38
rs778007668 Chr.10: 93612941-93612941 on Build GRCh38
rs752108634 Chr.10: 93612923-93612923 on Build GRCh38
rs764217729 Chr.10: 93613045-93613045 on Build GRCh38
rs541813651 Chr.10: 93612993-93612993 on Build GRCh38
rs767034843 Chr.10: 93612921-93612921 on Build GRCh38
rs557957487 Chr.10: 93612946-93612946 on Build GRCh38
rs758739604 Chr.10: 93612893-93612893 on Build GRCh38
rs201286677 Chr.10: 93612953-93612953 on Build GRCh38
rs201231586 Chr.10: 93613041-93613041 on Build GRCh38
rs749089806 Chr.10: 93612943-93612943 on Build GRCh38
rs367944390 Chr.10: 93612887-93612887 on Build GRCh38
rs375795507 Chr.10: 93613029-93613029 on Build GRCh38
rs138308024 Chr.10: 93613004-93613004 on Build GRCh38
rs992144807 Chr.10: 93613037-93613037 on Build GRCh38
rs776384738 Chr.10: 93612970-93612971 on Build GRCh38
rs1040611662 Chr.10: 93612966-93612966 on Build GRCh38
rs62642544 Chr.10: 93612928-93612928 on Build GRCh38
rs753799663 Chr.10: 93613046-93613046 on Build GRCh38
rs534132522 Chr.10: 93612972-93612972 on Build GRCh38
rs779992768 Chr.10: 93613023-93613023 on Build GRCh38
rs759082069 Chr.10: 93612855-93612855 on Build GRCh38
rs372464685 Chr.10: 93612932-93612932 on Build GRCh38
rs560963943 Chr.10: 93613056-93613056 on Build GRCh38
rs368555084 Chr.10: 93613013-93613013 on Build GRCh38
rs566501091 Chr.10: 93612869-93612869 on Build GRCh38
rs763733585 Chr.10: 93612889-93612889 on Build GRCh38
rs746549962 Chr.10: 93613025-93613025 on Build GRCh38
rs770812539 Chr.10: 93612941-93612941 on Build GRCh38
rs757099539 Chr.10: 93613008-93613008 on Build GRCh38
rs897789860 Chr.10: 93612968-93612968 on Build GRCh38
rs756324901 Chr.10: 93612936-93612936 on Build GRCh38
rs786200908 Chr.10: 93612982-93612983 on Build GRCh38
rs760729628 Chr.10: 93613043-93613043 on Build GRCh38
rs986337181 Chr.10: 93612929-93612929 on Build GRCh38
rs545034158 Chr.10: 93613031-93613031 on Build GRCh38
rs143619428 Chr.10: 93613040-93613040 on Build GRCh38
rs12781149 Chr.10: 93613007-93613007 on Build GRCh38
rs979715598 Chr.10: 93613017-93613017 on Build GRCh38
rs759933390 Chr.10: 93613038-93613038 on Build GRCh38
rs767007825 Chr.10: 93612885-93612885 on Build GRCh38
rs774245507 Chr.10: 93612849-93612849 on Build GRCh38
rs760190350 Chr.10: 93612962-93612962 on Build GRCh38
rs774984663 Chr.10: 93612954-93612954 on Build GRCh38
rs1131978 Chr.10: 93612977-93612977 on Build GRCh38
rs763675675 Chr.10: 93612967-93612967 on Build GRCh38
rs760127153 Chr.10: 93612990-93612990 on Build GRCh38
rs760329824 Chr.10: 93612868-93612868 on Build GRCh38

View Full Product Details