Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62648810-62649047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS5
Entrez Gene ID
80789
Gene Name
integrator complex subunit 5
Gene Aliases
INT5, KIAA1698
UniGene
Hs.458390
Gene Chromosome Location
Chr.11: 62646848-62653302 on Build GRCh38
Transcripts
NM_030628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4034841 c.1058G>A p.G353D Chr.11: 62649022-62649022 on Build GRCh38
6998093 c.1141C>T p.R381* Chr.11: 62648939-62648939 on Build GRCh38
7203176 c.1063G>C p.V355L Chr.11: 62649017-62649017 on Build GRCh38
4034840 c.1196C>A p.S399Y Chr.11: 62648884-62648884 on Build GRCh38
2039344 c.1230G>A p.L410L Chr.11: 62648850-62648850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767352618 Chr.11: 62648990-62648990 on Build GRCh38
rs551589391 Chr.11: 62648865-62648865 on Build GRCh38
rs750403759 Chr.11: 62648879-62648879 on Build GRCh38
rs746787451 Chr.11: 62648997-62648997 on Build GRCh38
rs887138568 Chr.11: 62648901-62648901 on Build GRCh38
rs773507778 Chr.11: 62648931-62648931 on Build GRCh38
rs763478352 Chr.11: 62648972-62648972 on Build GRCh38
rs773809662 Chr.11: 62648837-62648837 on Build GRCh38
rs756071716 Chr.11: 62648880-62648880 on Build GRCh38
rs764749943 Chr.11: 62648843-62648843 on Build GRCh38
rs373426235 Chr.11: 62648866-62648866 on Build GRCh38
rs780283106 Chr.11: 62648884-62648884 on Build GRCh38
rs375771026 Chr.11: 62648962-62648962 on Build GRCh38
rs777428024 Chr.11: 62648862-62648862 on Build GRCh38
rs377762006 Chr.11: 62648918-62648918 on Build GRCh38
rs757145863 Chr.11: 62648867-62648867 on Build GRCh38
rs761148413 Chr.11: 62648841-62648841 on Build GRCh38
rs749320731 Chr.11: 62648897-62648897 on Build GRCh38
rs774115672 Chr.11: 62648978-62648978 on Build GRCh38
rs754997482 Chr.11: 62648902-62648902 on Build GRCh38
rs771385752 Chr.11: 62648941-62648941 on Build GRCh38
rs767100627 Chr.11: 62648938-62648938 on Build GRCh38
rs968316905 Chr.11: 62648835-62648835 on Build GRCh38
rs748314315 Chr.11: 62648925-62648925 on Build GRCh38
rs146635467 Chr.11: 62648992-62648992 on Build GRCh38
rs940071618 Chr.11: 62649007-62649007 on Build GRCh38
rs144513848 Chr.11: 62648930-62648930 on Build GRCh38
rs370934303 Chr.11: 62648942-62648942 on Build GRCh38
rs762387237 Chr.11: 62648843-62648849 on Build GRCh38
rs774941934 Chr.11: 62648852-62648852 on Build GRCh38
rs762600944 Chr.11: 62648853-62648853 on Build GRCh38
rs775001114 Chr.11: 62648945-62648945 on Build GRCh38
rs761515526 Chr.11: 62648873-62648873 on Build GRCh38
rs767189491 Chr.11: 62648868-62648868 on Build GRCh38
rs756146061 Chr.11: 62648995-62648995 on Build GRCh38
rs761353889 Chr.11: 62648989-62648989 on Build GRCh38

View Full Product Details