Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45099350-45099623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMT1
Entrez Gene ID
4836
Gene Name
N-myristoyltransferase 1
Gene Aliases
NMT
UniGene
Hs.532790
Gene Chromosome Location
Chr.17: 45061291-45109016 on Build GRCh38
Transcripts
NM_021079.4 XM_011524858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513322 c.891G>A p.W297* Chr.17: 45099411-45099411 on Build GRCh38
4753854 c.910C>T p.R304W Chr.17: 45099430-45099430 on Build GRCh38
3518201 c.937C>T p.H313Y Chr.17: 45099457-45099457 on Build GRCh38
4450834 c.885-1G>A p.? Chr.17: 45099404-45099404 on Build GRCh38
5779597 c.918G>A p.L306L Chr.17: 45099438-45099438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897001493 Chr.17: 45099530-45099530 on Build GRCh38
rs2239923 Chr.17: 45099436-45099436 on Build GRCh38
rs760110087 Chr.17: 45099487-45099487 on Build GRCh38
rs748936607 Chr.17: 45099504-45099504 on Build GRCh38
rs749910282 Chr.17: 45099398-45099398 on Build GRCh38
rs779600514 Chr.17: 45099447-45099447 on Build GRCh38
rs895100652 Chr.17: 45099593-45099593 on Build GRCh38
rs559095378 Chr.17: 45099596-45099596 on Build GRCh38
rs769878243 Chr.17: 45099543-45099543 on Build GRCh38
rs1056950860 Chr.17: 45099441-45099445 on Build GRCh38
rs748011369 Chr.17: 45099388-45099388 on Build GRCh38
rs931168917 Chr.17: 45099590-45099590 on Build GRCh38
rs762308922 Chr.17: 45099539-45099540 on Build GRCh38
rs985161494 Chr.17: 45099438-45099438 on Build GRCh38
rs772701064 Chr.17: 45099486-45099486 on Build GRCh38
rs747584701 Chr.17: 45099528-45099528 on Build GRCh38
rs778267952 Chr.17: 45099527-45099527 on Build GRCh38
rs775617587 Chr.17: 45099553-45099553 on Build GRCh38
rs1048289857 Chr.17: 45099592-45099592 on Build GRCh38
rs200150620 Chr.17: 45099385-45099385 on Build GRCh38
rs762690021 Chr.17: 45099377-45099377 on Build GRCh38
rs190201082 Chr.17: 45099540-45099540 on Build GRCh38
rs202085249 Chr.17: 45099536-45099536 on Build GRCh38
rs775733922 Chr.17: 45099374-45099374 on Build GRCh38
rs758880042 Chr.17: 45099524-45099524 on Build GRCh38
rs767085625 Chr.17: 45099390-45099390 on Build GRCh38
rs566288330 Chr.17: 45099384-45099384 on Build GRCh38

View Full Product Details