Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135804908-135805133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCT
Entrez Gene ID
3938
Gene Name
lactase
Gene Aliases
LAC, LPH, LPH1
UniGene
Hs.551506
Gene Chromosome Location
Chr.2: 135787840-135837200 on Build GRCh38
Transcripts
NM_002299.3 XM_017004088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1399565 c.4224G>A p.T1408T Chr.2: 135805007-135805007 on Build GRCh38
3566783 c.4275C>T p.A1425A Chr.2: 135804956-135804956 on Build GRCh38
1256379 c.4190G>A p.R1397K Chr.2: 135805041-135805041 on Build GRCh38
5354476 c.4209C>T p.L1403L Chr.2: 135805022-135805022 on Build GRCh38
6249791 c.4274C>A p.A1425D Chr.2: 135804957-135804957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768376212 Chr.2: 135804940-135804940 on Build GRCh38
rs767744781 Chr.2: 135804986-135804986 on Build GRCh38
rs148051541 Chr.2: 135804958-135804958 on Build GRCh38
rs79686438 Chr.2: 135804956-135804956 on Build GRCh38
rs746550675 Chr.2: 135804932-135804932 on Build GRCh38
rs142685465 Chr.2: 135805002-135805002 on Build GRCh38
rs553173069 Chr.2: 135805087-135805087 on Build GRCh38
rs148790460 Chr.2: 135804933-135804933 on Build GRCh38
rs373989454 Chr.2: 135804971-135804971 on Build GRCh38
rs183913069 Chr.2: 135805046-135805046 on Build GRCh38
rs774926435 Chr.2: 135805073-135805073 on Build GRCh38
rs773638318 Chr.2: 135804962-135804962 on Build GRCh38
rs752925060 Chr.2: 135804992-135804992 on Build GRCh38
rs769491072 Chr.2: 135805088-135805088 on Build GRCh38
rs780893486 Chr.2: 135805077-135805077 on Build GRCh38
rs190978059 Chr.2: 135805023-135805023 on Build GRCh38
rs150523199 Chr.2: 135805012-135805012 on Build GRCh38
rs766901502 Chr.2: 135804977-135804977 on Build GRCh38
rs936013862 Chr.2: 135805110-135805110 on Build GRCh38
rs754487360 Chr.2: 135805076-135805076 on Build GRCh38
rs751586712 Chr.2: 135804981-135804981 on Build GRCh38
rs773406785 Chr.2: 135805097-135805097 on Build GRCh38
rs199901583 Chr.2: 135805007-135805007 on Build GRCh38
rs776196678 Chr.2: 135804952-135804952 on Build GRCh38
rs142467054 Chr.2: 135805066-135805066 on Build GRCh38
rs903218062 Chr.2: 135805106-135805106 on Build GRCh38
rs762207096 Chr.2: 135804961-135804961 on Build GRCh38
rs964897479 Chr.2: 135804957-135804957 on Build GRCh38
rs988407821 Chr.2: 135805033-135805033 on Build GRCh38
rs764886525 Chr.2: 135805008-135805008 on Build GRCh38

View Full Product Details