Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92466279-92466532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
UniGene
Hs.435655
Gene Chromosome Location
Chr.9: 92456205-92482562 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5120146 c.601+4A>C p.? Chr.9: 92466354-92466354 on Build GRCh38
4663802 c.445A>G p.T149A Chr.9: 92466514-92466514 on Build GRCh38
3982969 c.465G>C p.L155L Chr.9: 92466494-92466494 on Build GRCh38
6573294 c.538G>A p.E180K Chr.9: 92466421-92466421 on Build GRCh38
6232074 c.510C>A p.P170P Chr.9: 92466449-92466449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41278697 Chr.9: 92466418-92466418 on Build GRCh38
rs371768627 Chr.9: 92466501-92466501 on Build GRCh38
rs565810963 Chr.9: 92466491-92466491 on Build GRCh38
rs750143544 Chr.9: 92466476-92466476 on Build GRCh38
rs747550942 Chr.9: 92466461-92466461 on Build GRCh38
rs931618671 Chr.9: 92466379-92466379 on Build GRCh38
rs369326728 Chr.9: 92466453-92466453 on Build GRCh38
rs774799220 Chr.9: 92466312-92466315 on Build GRCh38
rs921706454 Chr.9: 92466366-92466366 on Build GRCh38
rs762136374 Chr.9: 92466314-92466315 on Build GRCh38
rs776917242 Chr.9: 92466478-92466478 on Build GRCh38
rs776877419 Chr.9: 92466342-92466342 on Build GRCh38
rs146775001 Chr.9: 92466394-92466394 on Build GRCh38
rs41278699 Chr.9: 92466494-92466494 on Build GRCh38
rs966554052 Chr.9: 92466450-92466450 on Build GRCh38
rs778643481 Chr.9: 92466445-92466445 on Build GRCh38
rs769055070 Chr.9: 92466330-92466330 on Build GRCh38
rs757923286 Chr.9: 92466454-92466454 on Build GRCh38
rs746176926 Chr.9: 92466444-92466444 on Build GRCh38
rs187543149 Chr.9: 92466321-92466321 on Build GRCh38
rs1013763853 Chr.9: 92466457-92466457 on Build GRCh38
rs756921551 Chr.9: 92466439-92466439 on Build GRCh38
rs747321863 Chr.9: 92466329-92466329 on Build GRCh38
rs143358512 Chr.9: 92466502-92466502 on Build GRCh38
rs772512883 Chr.9: 92466312-92466312 on Build GRCh38
rs755951081 Chr.9: 92466414-92466414 on Build GRCh38
rs767126722 Chr.9: 92466436-92466438 on Build GRCh38
rs753528217 Chr.9: 92466433-92466433 on Build GRCh38
rs748558453 Chr.9: 92466480-92466480 on Build GRCh38
rs144954352 Chr.9: 92466365-92466365 on Build GRCh38
rs368329214 Chr.9: 92466469-92466469 on Build GRCh38
rs765345253 Chr.9: 92466372-92466372 on Build GRCh38
rs371262202 Chr.9: 92466403-92466403 on Build GRCh38
rs769057452 Chr.9: 92466470-92466470 on Build GRCh38
rs1006311203 Chr.9: 92466353-92466353 on Build GRCh38
rs763128075 Chr.9: 92466386-92466386 on Build GRCh38
rs780513757 Chr.9: 92466458-92466458 on Build GRCh38
rs41278695 Chr.9: 92466381-92466381 on Build GRCh38
rs374381931 Chr.9: 92466452-92466452 on Build GRCh38

View Full Product Details