Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201807839-201808064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156572 c.159G>T p.L53F Chr.2: 201807896-201807896 on Build GRCh38
6156574 c.129C>A p.S43R Chr.2: 201807866-201807866 on Build GRCh38
319385 c.196G>T p.V66F Chr.2: 201807933-201807933 on Build GRCh38
6156573 c.312G>T p.L104F Chr.2: 201807896-201807896 on Build GRCh38
476757 c.201C>A p.Y67* Chr.2: 201807938-201807938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148612179 Chr.2: 201808006-201808006 on Build GRCh38
rs750372686 Chr.2: 201807990-201807990 on Build GRCh38
rs751194818 Chr.2: 201807975-201807975 on Build GRCh38
rs769653908 Chr.2: 201807879-201807879 on Build GRCh38
rs149189465 Chr.2: 201807928-201807928 on Build GRCh38
rs754267888 Chr.2: 201807926-201807926 on Build GRCh38
rs373249831 Chr.2: 201807996-201807996 on Build GRCh38
rs199758419 Chr.2: 201807909-201807909 on Build GRCh38
rs545008145 Chr.2: 201807997-201807998 on Build GRCh38
rs761901115 Chr.2: 201807913-201807913 on Build GRCh38
rs780051899 Chr.2: 201807883-201807883 on Build GRCh38
rs955385484 Chr.2: 201807889-201807889 on Build GRCh38
rs79681158 Chr.2: 201808037-201808037 on Build GRCh38
rs960275137 Chr.2: 201807995-201807995 on Build GRCh38
rs768959454 Chr.2: 201807888-201807888 on Build GRCh38
rs565287827 Chr.2: 201808034-201808034 on Build GRCh38
rs540681761 Chr.2: 201808015-201808015 on Build GRCh38
rs766721567 Chr.2: 201807924-201807924 on Build GRCh38
rs1042852430 Chr.2: 201808022-201808022 on Build GRCh38
rs772172729 Chr.2: 201807915-201807915 on Build GRCh38
rs756717654 Chr.2: 201807976-201807976 on Build GRCh38
rs992412291 Chr.2: 201807964-201807964 on Build GRCh38
rs535163624 Chr.2: 201807911-201807911 on Build GRCh38
rs748284126 Chr.2: 201807972-201807972 on Build GRCh38
rs780861693 Chr.2: 201807981-201807981 on Build GRCh38
rs374756467 Chr.2: 201808003-201808003 on Build GRCh38
rs777532856 Chr.2: 201807964-201807965 on Build GRCh38
rs779959825 Chr.2: 201808002-201808002 on Build GRCh38
rs763857342 Chr.2: 201807929-201807929 on Build GRCh38
rs982783078 Chr.2: 201807890-201807890 on Build GRCh38
rs761187271 Chr.2: 201807919-201807919 on Build GRCh38
rs776836300 Chr.2: 201807961-201807961 on Build GRCh38
rs138441846 Chr.2: 201807891-201807891 on Build GRCh38
rs997863043 Chr.2: 201807870-201807870 on Build GRCh38
rs77557988 Chr.2: 201807887-201807887 on Build GRCh38

View Full Product Details