Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2280850-2281119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
UniGene
Hs.26630
Gene Chromosome Location
Chr.16: 2275878-2340746 on Build GRCh38
Transcripts
NM_001089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589711 c.4353C>T p.V1451V Chr.16: 2281033-2281033 on Build GRCh38
6884933 c.4327G>C p.G1443R Chr.16: 2281059-2281059 on Build GRCh38
5606285 c.4319C>T p.A1440V Chr.16: 2281067-2281067 on Build GRCh38
252311 c.4354G>A p.G1452R Chr.16: 2281032-2281032 on Build GRCh38
3817556 c.4287G>A p.L1429L Chr.16: 2281099-2281099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978053004 Chr.16: 2280883-2280883 on Build GRCh38
rs760644085 Chr.16: 2281055-2281055 on Build GRCh38
rs1029584079 Chr.16: 2281015-2281015 on Build GRCh38
rs767302159 Chr.16: 2281033-2281033 on Build GRCh38
rs755504528 Chr.16: 2281095-2281095 on Build GRCh38
rs189737924 Chr.16: 2281096-2281096 on Build GRCh38
rs34902969 Chr.16: 2281095-2281096 on Build GRCh38
rs769129570 Chr.16: 2280995-2280995 on Build GRCh38
rs538210968 Chr.16: 2281060-2281060 on Build GRCh38
rs747556947 Chr.16: 2280992-2280992 on Build GRCh38
rs376588751 Chr.16: 2280991-2280991 on Build GRCh38
rs375783245 Chr.16: 2281032-2281032 on Build GRCh38
rs576334134 Chr.16: 2281011-2281011 on Build GRCh38
rs1048803367 Chr.16: 2280900-2280900 on Build GRCh38
rs889654694 Chr.16: 2280975-2280975 on Build GRCh38
rs536127710 Chr.16: 2280893-2280893 on Build GRCh38
rs372848304 Chr.16: 2281013-2281013 on Build GRCh38
rs1015185214 Chr.16: 2280931-2280931 on Build GRCh38
rs556151715 Chr.16: 2280920-2280920 on Build GRCh38
rs770589758 Chr.16: 2281025-2281025 on Build GRCh38
rs199852776 Chr.16: 2281081-2281081 on Build GRCh38
rs775345113 Chr.16: 2281044-2281044 on Build GRCh38
rs772765938 Chr.16: 2280997-2280997 on Build GRCh38
rs569956903 Chr.16: 2280980-2280980 on Build GRCh38
rs750703859 Chr.16: 2281083-2281083 on Build GRCh38
rs747392070 Chr.16: 2281093-2281093 on Build GRCh38
rs780579884 Chr.16: 2281087-2281087 on Build GRCh38
rs397518429 Chr.16: 2281064-2281064 on Build GRCh38
rs933898218 Chr.16: 2280961-2280961 on Build GRCh38
rs1033077802 Chr.16: 2281057-2281057 on Build GRCh38
rs772585636 Chr.16: 2280984-2280984 on Build GRCh38
rs765505615 Chr.16: 2281082-2281082 on Build GRCh38
rs1050037960 Chr.16: 2280963-2280963 on Build GRCh38
rs937026510 Chr.16: 2280887-2280887 on Build GRCh38
rs896878101 Chr.16: 2281029-2281029 on Build GRCh38
rs572167727 Chr.16: 2281085-2281085 on Build GRCh38
rs887548774 Chr.16: 2280901-2280901 on Build GRCh38
rs764001106 Chr.16: 2281059-2281059 on Build GRCh38
rs770842336 Chr.16: 2280916-2280916 on Build GRCh38

View Full Product Details