Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23757523-23757748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
-
UniGene
Hs.178589
Gene Chromosome Location
Chr.16: 23754627-23758951 on Build GRCh38
Transcripts
NM_022097.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957357 c.540C>A p.S180S Chr.16: 23757532-23757532 on Build GRCh38
1248261 c.563A>G p.Q188R Chr.16: 23757555-23757555 on Build GRCh38
3711985 c.577C>T p.R193W Chr.16: 23757569-23757569 on Build GRCh38
5913748 c.578G>A p.R193Q Chr.16: 23757570-23757570 on Build GRCh38
5909490 c.540C>T p.S180S Chr.16: 23757532-23757532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747054858 Chr.16: 23757602-23757602 on Build GRCh38
rs1013305043 Chr.16: 23757712-23757712 on Build GRCh38
rs987941968 Chr.16: 23757562-23757562 on Build GRCh38
rs762723340 Chr.16: 23757568-23757568 on Build GRCh38
rs762385078 Chr.16: 23757621-23757621 on Build GRCh38
rs755849123 Chr.16: 23757579-23757579 on Build GRCh38
rs944966416 Chr.16: 23757594-23757594 on Build GRCh38
rs751008002 Chr.16: 23757570-23757570 on Build GRCh38
rs941223791 Chr.16: 23757654-23757654 on Build GRCh38
rs149423884 Chr.16: 23757588-23757588 on Build GRCh38
rs566201714 Chr.16: 23757688-23757688 on Build GRCh38
rs777550065 Chr.16: 23757585-23757585 on Build GRCh38
rs1043323702 Chr.16: 23757661-23757661 on Build GRCh38
rs777261472 Chr.16: 23757547-23757547 on Build GRCh38
rs780991203 Chr.16: 23757575-23757575 on Build GRCh38
rs754769864 Chr.16: 23757586-23757586 on Build GRCh38
rs1050553557 Chr.16: 23757637-23757637 on Build GRCh38
rs558686269 Chr.16: 23757653-23757653 on Build GRCh38
rs534098372 Chr.16: 23757643-23757643 on Build GRCh38
rs923403762 Chr.16: 23757624-23757624 on Build GRCh38
rs903509361 Chr.16: 23757662-23757662 on Build GRCh38
rs758234304 Chr.16: 23757649-23757649 on Build GRCh38
rs149498782 Chr.16: 23757548-23757548 on Build GRCh38
rs768838800 Chr.16: 23757615-23757615 on Build GRCh38
rs61731149 Chr.16: 23757554-23757554 on Build GRCh38
rs371343275 Chr.16: 23757627-23757627 on Build GRCh38
rs1049154584 Chr.16: 23757648-23757648 on Build GRCh38
rs144857717 Chr.16: 23757645-23757645 on Build GRCh38
rs780374025 Chr.16: 23757595-23757595 on Build GRCh38
rs770240666 Chr.16: 23757622-23757622 on Build GRCh38
rs999241225 Chr.16: 23757668-23757668 on Build GRCh38
rs16972893 Chr.16: 23757587-23757587 on Build GRCh38
rs755012244 Chr.16: 23757689-23757689 on Build GRCh38
rs754560117 Chr.16: 23757572-23757572 on Build GRCh38
rs765935235 Chr.16: 23757569-23757569 on Build GRCh38
rs780647388 Chr.16: 23757669-23757669 on Build GRCh38
rs776943808 Chr.16: 23757617-23757617 on Build GRCh38

View Full Product Details