Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27658401-27658626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
UniGene
Hs.502182
Gene Chromosome Location
Chr.11: 27654893-27722058 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Gene Symbol
BDNF-AS
Entrez Gene ID
497258
Gene Name
BDNF antisense RNA
Gene Aliases
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
UniGene
Hs.675323
Gene Chromosome Location
Chr.11: 27506852-27698171 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4869018 c.196G>A p.A66T Chr.11: 27658414-27658414 on Build GRCh38
7210256 c.156G>A p.V52V Chr.11: 27658454-27658454 on Build GRCh38
3446587 c.271G>A p.V91I Chr.11: 27658540-27658540 on Build GRCh38
4032242 c.111A>C p.Q37H Chr.11: 27658478-27658478 on Build GRCh38
7109087 c.77C>T p.S26L Chr.11: 27658533-27658533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943009748 Chr.11: 27658451-27658451 on Build GRCh38
rs780865517 Chr.11: 27658580-27658580 on Build GRCh38
rs764254110 Chr.11: 27658495-27658495 on Build GRCh38
rs779925777 Chr.11: 27658546-27658546 on Build GRCh38
rs759834365 Chr.11: 27658456-27658456 on Build GRCh38
rs1037807065 Chr.11: 27658458-27658458 on Build GRCh38
rs146354977 Chr.11: 27658429-27658429 on Build GRCh38
rs982182462 Chr.11: 27658569-27658569 on Build GRCh38
rs990506363 Chr.11: 27658485-27658485 on Build GRCh38
rs551445385 Chr.11: 27658592-27658592 on Build GRCh38
rs374752426 Chr.11: 27658454-27658454 on Build GRCh38
rs760902255 Chr.11: 27658490-27658490 on Build GRCh38
rs8192466 Chr.11: 27658560-27658560 on Build GRCh38
rs267602837 Chr.11: 27658486-27658486 on Build GRCh38
rs747701929 Chr.11: 27658430-27658430 on Build GRCh38
rs768158526 Chr.11: 27658421-27658421 on Build GRCh38
rs750397219 Chr.11: 27658516-27658516 on Build GRCh38
rs763357890 Chr.11: 27658452-27658452 on Build GRCh38
rs771341699 Chr.11: 27658453-27658453 on Build GRCh38
rs920340884 Chr.11: 27658484-27658484 on Build GRCh38
rs866172975 Chr.11: 27658533-27658533 on Build GRCh38
rs758200229 Chr.11: 27658526-27658526 on Build GRCh38
rs767681787 Chr.11: 27658473-27658473 on Build GRCh38
rs769259549 Chr.11: 27658432-27658432 on Build GRCh38
rs868827437 Chr.11: 27658507-27658507 on Build GRCh38
rs773773442 Chr.11: 27658444-27658444 on Build GRCh38

View Full Product Details