Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31845402-31845645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP15
Entrez Gene ID
128853
Gene Name
dual specificity phosphatase 15
Gene Aliases
C20orf57, VHY
UniGene
Hs.434108
Gene Chromosome Location
Chr.20: 31845578-31870744 on Build GRCh38
Transcripts
XM_017027656.1 NM_004118.3

Gene Symbol
FOXS1
Entrez Gene ID
2307
Gene Name
forkhead box S1
Gene Aliases
FKHL18, FREAC10
UniGene
Hs.516971
Gene Chromosome Location
Chr.20: 31844300-31845617 on Build GRCh38
Transcripts
XM_017027656.1 NM_004118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681590 c.111G>C p.Q37H Chr.20: 31845432-31845432 on Build GRCh38
5907054 c.58C>T p.P20S Chr.20: 31845485-31845485 on Build GRCh38
311275 c.31G>A p.A11T Chr.20: 31845512-31845512 on Build GRCh38
7161053 c.132C>G p.I44M Chr.20: 31845411-31845411 on Build GRCh38
3911057 c.90C>T p.A30A Chr.20: 31845453-31845453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142596973 Chr.20: 31845521-31845521 on Build GRCh38
rs777899171 Chr.20: 31845468-31845468 on Build GRCh38
rs550780074 Chr.20: 31845622-31845622 on Build GRCh38
rs765446653 Chr.20: 31845447-31845447 on Build GRCh38
rs556551324 Chr.20: 31845603-31845603 on Build GRCh38
rs766343792 Chr.20: 31845438-31845438 on Build GRCh38
rs755181622 Chr.20: 31845444-31845444 on Build GRCh38
rs546382925 Chr.20: 31845535-31845535 on Build GRCh38
rs970549542 Chr.20: 31845588-31845588 on Build GRCh38
rs757212795 Chr.20: 31845477-31845477 on Build GRCh38
rs775193708 Chr.20: 31845503-31845503 on Build GRCh38
rs757146374 Chr.20: 31845563-31845563 on Build GRCh38
rs751360729 Chr.20: 31845550-31845550 on Build GRCh38
rs759652903 Chr.20: 31845525-31845525 on Build GRCh38
rs868579840 Chr.20: 31845454-31845454 on Build GRCh38
rs375219720 Chr.20: 31845505-31845505 on Build GRCh38
rs564111331 Chr.20: 31845522-31845522 on Build GRCh38
rs945429498 Chr.20: 31845625-31845625 on Build GRCh38
rs144561926 Chr.20: 31845500-31845500 on Build GRCh38
rs752495717 Chr.20: 31845448-31845448 on Build GRCh38
rs758320257 Chr.20: 31845539-31845539 on Build GRCh38
rs780598711 Chr.20: 31845489-31845489 on Build GRCh38
rs781126376 Chr.20: 31845583-31845583 on Build GRCh38
rs529098872 Chr.20: 31845617-31845617 on Build GRCh38
rs563064487 Chr.20: 31845439-31845439 on Build GRCh38
rs760782281 Chr.20: 31845431-31845431 on Build GRCh38
rs981991405 Chr.20: 31845602-31845602 on Build GRCh38
rs368093781 Chr.20: 31845513-31845513 on Build GRCh38
rs937980747 Chr.20: 31845607-31845607 on Build GRCh38
rs890073032 Chr.20: 31845527-31845527 on Build GRCh38
rs145658539 Chr.20: 31845512-31845512 on Build GRCh38
rs745426844 Chr.20: 31845590-31845590 on Build GRCh38
rs142041083 Chr.20: 31845494-31845494 on Build GRCh38
rs6121246 Chr.20: 31845549-31845549 on Build GRCh38
rs991999994 Chr.20: 31845589-31845589 on Build GRCh38
rs772392827 Chr.20: 31845507-31845507 on Build GRCh38
rs528192984 Chr.20: 31845534-31845534 on Build GRCh38
rs746922689 Chr.20: 31845471-31845471 on Build GRCh38
rs773702165 Chr.20: 31845430-31845430 on Build GRCh38
rs947446479 Chr.20: 31845605-31845605 on Build GRCh38
rs915260057 Chr.20: 31845620-31845620 on Build GRCh38
rs758267106 Chr.20: 31845456-31845456 on Build GRCh38
rs768069261 Chr.20: 31845423-31845423 on Build GRCh38

View Full Product Details