Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52461794-52462053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2
Entrez Gene ID
26586
Gene Name
cytoskeleton associated protein 2
Gene Aliases
LB1, TMAP, se20-10
UniGene
Hs.444028
Gene Chromosome Location
Chr.13: 52455360-52476631 on Build GRCh38
Transcripts
NM_001098525.2 NM_001286686.1 NM_001286687.1 NM_018204.4 XM_005266344.4 XM_011535043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1943436 c.1017C>T p.P339P Chr.13: 52461843-52461843 on Build GRCh38
1943437 c.1027C>T p.R343* Chr.13: 52461853-52461853 on Build GRCh38
5121468 c.437_1305+420>25 p.? Chr.13: 52461263-52462987 on Build GRCh38
6074526 c.1097G>C p.R366T Chr.13: 52461923-52461923 on Build GRCh38
6351733 c.999G>A p.L333L Chr.13: 52461825-52461825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769189054 Chr.13: 52461862-52461862 on Build GRCh38
rs375430927 Chr.13: 52461909-52461909 on Build GRCh38
rs3809374 Chr.13: 52461825-52461825 on Build GRCh38
rs775165143 Chr.13: 52461863-52461863 on Build GRCh38
rs760539520 Chr.13: 52461843-52461843 on Build GRCh38
rs776766061 Chr.13: 52461844-52461844 on Build GRCh38
rs189328880 Chr.13: 52461946-52461946 on Build GRCh38
rs762531256 Chr.13: 52461872-52461872 on Build GRCh38
rs752872038 Chr.13: 52461928-52461928 on Build GRCh38
rs181351481 Chr.13: 52461999-52461999 on Build GRCh38
rs750237853 Chr.13: 52461928-52461929 on Build GRCh38
rs745828219 Chr.13: 52461962-52461962 on Build GRCh38
rs538180566 Chr.13: 52461898-52461898 on Build GRCh38
rs777413181 Chr.13: 52461958-52461958 on Build GRCh38
rs778122467 Chr.13: 52461935-52461935 on Build GRCh38
rs1049625668 Chr.13: 52461893-52461893 on Build GRCh38
rs757129009 Chr.13: 52461938-52461938 on Build GRCh38
rs1046911203 Chr.13: 52461945-52461945 on Build GRCh38
rs758517766 Chr.13: 52461933-52461933 on Build GRCh38
rs750786244 Chr.13: 52461895-52461895 on Build GRCh38
rs754349566 Chr.13: 52461911-52461911 on Build GRCh38
rs370566115 Chr.13: 52462007-52462007 on Build GRCh38
rs943796314 Chr.13: 52461978-52461978 on Build GRCh38
rs929827367 Chr.13: 52461941-52461941 on Build GRCh38
rs763041884 Chr.13: 52461936-52461937 on Build GRCh38
rs748899714 Chr.13: 52461977-52461977 on Build GRCh38
rs778882235 Chr.13: 52461915-52461915 on Build GRCh38
rs769294913 Chr.13: 52461965-52461965 on Build GRCh38
rs9536083 Chr.13: 52462028-52462028 on Build GRCh38
rs201553403 Chr.13: 52461881-52461881 on Build GRCh38
rs759056863 Chr.13: 52461860-52461860 on Build GRCh38
rs141769986 Chr.13: 52461914-52461914 on Build GRCh38
rs761678480 Chr.13: 52461926-52461927 on Build GRCh38
rs571468996 Chr.13: 52461969-52461969 on Build GRCh38
rs746785764 Chr.13: 52461937-52461937 on Build GRCh38
rs752237934 Chr.13: 52461854-52461854 on Build GRCh38
rs755829774 Chr.13: 52461931-52461931 on Build GRCh38

View Full Product Details