Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133578951-133579190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM163B
Entrez Gene ID
642968
Gene Name
family with sequence similarity 163 member B
Gene Aliases
C9orf166
UniGene
Hs.173134
Gene Chromosome Location
Chr.9: 133577075-133609953 on Build GRCh38
Transcripts
NM_001080515.2 XM_005272203.4 XM_005272204.4 XM_006717233.3 XM_011518912.2 XM_011518913.2 XM_017015017.1 XM_017015018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3926381 c.381C>G p.S127R Chr.9: 133579142-133579142 on Build GRCh38
6335822 c.397C>T p.L133L Chr.9: 133579126-133579126 on Build GRCh38
4545461 c.378G>A p.V126V Chr.9: 133579145-133579145 on Build GRCh38
4545760 c.385G>A p.E129K Chr.9: 133579138-133579138 on Build GRCh38
6367068 c.364C>T p.L122F Chr.9: 133579159-133579159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567087803 Chr.9: 133579136-133579136 on Build GRCh38
rs757130732 Chr.9: 133579172-133579172 on Build GRCh38
rs75377207 Chr.9: 133579109-133579109 on Build GRCh38
rs778935577 Chr.9: 133579010-133579010 on Build GRCh38
rs771220753 Chr.9: 133579027-133579028 on Build GRCh38
rs1027102743 Chr.9: 133579030-133579030 on Build GRCh38
rs770755862 Chr.9: 133579055-133579055 on Build GRCh38
rs773430083 Chr.9: 133579162-133579162 on Build GRCh38
rs1029207648 Chr.9: 133579161-133579161 on Build GRCh38
rs756929157 Chr.9: 133579116-133579116 on Build GRCh38
rs761996588 Chr.9: 133579091-133579091 on Build GRCh38
rs113641726 Chr.9: 133579094-133579094 on Build GRCh38
rs534382587 Chr.9: 133579163-133579163 on Build GRCh38
rs555269190 Chr.9: 133579117-133579117 on Build GRCh38
rs745401450 Chr.9: 133579068-133579068 on Build GRCh38
rs371125485 Chr.9: 133579120-133579120 on Build GRCh38
rs550586461 Chr.9: 133579081-133579081 on Build GRCh38
rs776624508 Chr.9: 133579164-133579164 on Build GRCh38
rs763970379 Chr.9: 133579166-133579166 on Build GRCh38
rs765019785 Chr.9: 133579104-133579104 on Build GRCh38
rs746205354 Chr.9: 133579168-133579168 on Build GRCh38
rs571577864 Chr.9: 133579113-133579113 on Build GRCh38
rs777497301 Chr.9: 133579052-133579052 on Build GRCh38
rs927067707 Chr.9: 133579102-133579102 on Build GRCh38
rs954229404 Chr.9: 133579044-133579044 on Build GRCh38
rs752379167 Chr.9: 133579012-133579012 on Build GRCh38
rs11507482 Chr.9: 133579118-133579118 on Build GRCh38
rs374236190 Chr.9: 133579147-133579147 on Build GRCh38
rs887708270 Chr.9: 133579013-133579013 on Build GRCh38
rs532154002 Chr.9: 133579080-133579080 on Build GRCh38
rs1017285546 Chr.9: 133579006-133579006 on Build GRCh38
rs754873379 Chr.9: 133579007-133579007 on Build GRCh38
rs965207304 Chr.9: 133579080-133579081 on Build GRCh38
rs1056450854 Chr.9: 133579167-133579167 on Build GRCh38
rs113641726 Chr.9: 133579094-133579094 on Build GRCh38
rs200727742 Chr.9: 133579054-133579054 on Build GRCh38
rs1004732995 Chr.9: 133579021-133579021 on Build GRCh38
rs1014775368 Chr.9: 133579023-133579023 on Build GRCh38
rs758327003 Chr.9: 133579108-133579108 on Build GRCh38
rs762916999 Chr.9: 133579165-133579165 on Build GRCh38
rs749785225 Chr.9: 133579132-133579132 on Build GRCh38
rs774632311 Chr.9: 133579085-133579085 on Build GRCh38
rs772227656 Chr.9: 133579148-133579148 on Build GRCh38
rs1008577421 Chr.9: 133578994-133578994 on Build GRCh38
rs752677803 Chr.9: 133579106-133579106 on Build GRCh38
rs890085739 Chr.9: 133578970-133578970 on Build GRCh38
rs755651229 Chr.9: 133579119-133579119 on Build GRCh38
rs780818543 Chr.9: 133579065-133579065 on Build GRCh38
rs751350352 Chr.9: 133579171-133579171 on Build GRCh38
rs901705165 Chr.9: 133579028-133579028 on Build GRCh38
rs890398593 Chr.9: 133579114-133579114 on Build GRCh38
rs760760094 Chr.9: 133579096-133579096 on Build GRCh38
rs11507480 Chr.9: 133579112-133579112 on Build GRCh38
rs781724280 Chr.9: 133579112-133579113 on Build GRCh38
rs1038664062 Chr.9: 133579159-133579159 on Build GRCh38
rs773231301 Chr.9: 133579095-133579095 on Build GRCh38
rs747507929 Chr.9: 133579005-133579006 on Build GRCh38
rs796561410 Chr.9: 133579031-133579031 on Build GRCh38
rs758219287 Chr.9: 133579045-133579045 on Build GRCh38

View Full Product Details