Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50325624-50325849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5S
Entrez Gene ID
27109
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases
ATPW, FB, HSU79253
UniGene
Hs.438489
Gene Chromosome Location
Chr.14: 50311752-50326715 on Build GRCh38
Transcripts
NM_001003803.2 NM_001003805.2 NR_033761.1 XM_005267537.3 XM_011536649.2 XM_011536651.2 XM_011536652.1 XM_011536653.2 XM_011536655.2 XM_011536657.2 XM_017021220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4333134 c.475G>C p.D159H Chr.14: 50325634-50325634 on Build GRCh38
4333132 c.559G>C p.D187H Chr.14: 50325634-50325634 on Build GRCh38
6607982 c.615G>A p.L205L Chr.14: 50325690-50325690 on Build GRCh38
5509205 c.469T>C p.L157L Chr.14: 50325628-50325628 on Build GRCh38
956038 c.583G>T p.E195* Chr.14: 50325658-50325658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772523439 Chr.14: 50325775-50325775 on Build GRCh38
rs576036774 Chr.14: 50325819-50325819 on Build GRCh38
rs774381451 Chr.14: 50325669-50325669 on Build GRCh38
rs766889535 Chr.14: 50325751-50325751 on Build GRCh38
rs748784549 Chr.14: 50325658-50325658 on Build GRCh38
rs752003281 Chr.14: 50325754-50325754 on Build GRCh38
rs765359072 Chr.14: 50325719-50325719 on Build GRCh38
rs922227062 Chr.14: 50325667-50325667 on Build GRCh38
rs1019197167 Chr.14: 50325746-50325746 on Build GRCh38
rs755299956 Chr.14: 50325768-50325768 on Build GRCh38
rs935955003 Chr.14: 50325756-50325756 on Build GRCh38
rs112164857 Chr.14: 50325664-50325664 on Build GRCh38
rs369428246 Chr.14: 50325714-50325714 on Build GRCh38
rs771834416 Chr.14: 50325681-50325681 on Build GRCh38
rs144057879 Chr.14: 50325732-50325732 on Build GRCh38
rs775147044 Chr.14: 50325692-50325692 on Build GRCh38
rs745672927 Chr.14: 50325670-50325670 on Build GRCh38
rs140298574 Chr.14: 50325677-50325677 on Build GRCh38

View Full Product Details