Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38702864-38703089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_011537951.2 XM_011537952.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674404 c.950C>A p.T317K Chr.12: 38702886-38702886 on Build GRCh38
6271493 c.930C>A p.F310L Chr.12: 38702906-38702906 on Build GRCh38
5508135 c.934G>A p.V312M Chr.12: 38702902-38702902 on Build GRCh38
4600237 c.952G>A p.A318T Chr.12: 38702884-38702884 on Build GRCh38
5923960 c.943G>A p.D315N Chr.12: 38702893-38702893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007341681 Chr.12: 38702991-38702991 on Build GRCh38
rs766429653 Chr.12: 38702990-38702990 on Build GRCh38
rs186928511 Chr.12: 38702942-38702942 on Build GRCh38
rs954394403 Chr.12: 38703037-38703037 on Build GRCh38
rs985532416 Chr.12: 38703054-38703054 on Build GRCh38
rs143462849 Chr.12: 38702919-38702919 on Build GRCh38
rs929906941 Chr.12: 38702933-38702933 on Build GRCh38
rs542924565 Chr.12: 38702918-38702918 on Build GRCh38
rs369650655 Chr.12: 38702967-38702967 on Build GRCh38
rs967511673 Chr.12: 38703049-38703049 on Build GRCh38
rs574389197 Chr.12: 38702915-38702915 on Build GRCh38
rs756910756 Chr.12: 38702971-38702971 on Build GRCh38
rs545662606 Chr.12: 38702976-38702976 on Build GRCh38
rs866699204 Chr.12: 38702960-38702961 on Build GRCh38
rs1018268336 Chr.12: 38703011-38703011 on Build GRCh38
rs111493425 Chr.12: 38702923-38702923 on Build GRCh38
rs763904688 Chr.12: 38702960-38702960 on Build GRCh38
rs1020483429 Chr.12: 38703026-38703026 on Build GRCh38
rs893218774 Chr.12: 38703019-38703019 on Build GRCh38
rs752297848 Chr.12: 38702949-38702949 on Build GRCh38
rs780216622 Chr.12: 38702910-38702910 on Build GRCh38
rs377183448 Chr.12: 38702927-38702927 on Build GRCh38
rs565355199 Chr.12: 38702988-38702988 on Build GRCh38
rs199569558 Chr.12: 38702896-38702896 on Build GRCh38
rs757415938 Chr.12: 38702901-38702901 on Build GRCh38
rs555732255 Chr.12: 38703060-38703060 on Build GRCh38
rs765033881 Chr.12: 38702947-38702947 on Build GRCh38
rs377245797 Chr.12: 38702943-38702943 on Build GRCh38
rs192238630 Chr.12: 38703025-38703025 on Build GRCh38
rs762519540 Chr.12: 38702953-38702953 on Build GRCh38
rs1038168295 Chr.12: 38702917-38702917 on Build GRCh38
rs1027155869 Chr.12: 38703035-38703035 on Build GRCh38

View Full Product Details