Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102027969-102028232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX1
Entrez Gene ID
1523
Gene Name
cut like homeobox 1
Gene Aliases
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
UniGene
Hs.191482
Gene Chromosome Location
Chr.7: 101815904-102283958 on Build GRCh38
Transcripts
NM_001202543.1 NM_001202544.2 NM_001202545.2 NM_001202546.2 NM_001913.4 NM_181500.3 NM_181552.3 XM_005250150.2 XM_005250151.2 XM_005250154.3 XM_006715854.2 XM_006715855.1 XM_011515823.2 XM_011515825.1 XM_017011759.1 XM_017011760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280678 c.175-3C>A p.? Chr.7: 102028095-102028095 on Build GRCh38
6638278 c.222+2T>C p.? Chr.7: 102028147-102028147 on Build GRCh38
6280677 c.142-3C>A p.? Chr.7: 102028095-102028095 on Build GRCh38
6638318 c.189+1G>T p.? Chr.7: 102028146-102028146 on Build GRCh38
6638277 c.189+2T>C p.? Chr.7: 102028147-102028147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765166573 Chr.7: 102028063-102028063 on Build GRCh38
rs4729769 Chr.7: 102028014-102028014 on Build GRCh38
rs757627662 Chr.7: 102028154-102028154 on Build GRCh38
rs531840329 Chr.7: 102028025-102028025 on Build GRCh38
rs989872151 Chr.7: 102028131-102028131 on Build GRCh38
rs182471866 Chr.7: 102027993-102027993 on Build GRCh38
rs4727517 Chr.7: 102028022-102028022 on Build GRCh38
rs550059780 Chr.7: 102028049-102028049 on Build GRCh38
rs754267064 Chr.7: 102028121-102028121 on Build GRCh38
rs754817839 Chr.7: 102028056-102028058 on Build GRCh38
rs779319768 Chr.7: 102028181-102028181 on Build GRCh38
rs780147486 Chr.7: 102028194-102028194 on Build GRCh38
rs377542184 Chr.7: 102028059-102028059 on Build GRCh38
rs751229655 Chr.7: 102028002-102028002 on Build GRCh38
rs143267032 Chr.7: 102028117-102028117 on Build GRCh38
rs1004452607 Chr.7: 102028037-102028037 on Build GRCh38
rs773165435 Chr.7: 102028068-102028068 on Build GRCh38
rs547996935 Chr.7: 102028185-102028185 on Build GRCh38
rs762396295 Chr.7: 102028080-102028080 on Build GRCh38
rs376238545 Chr.7: 102028186-102028186 on Build GRCh38
rs200849897 Chr.7: 102028083-102028083 on Build GRCh38
rs778507488 Chr.7: 102028078-102028078 on Build GRCh38
rs374397164 Chr.7: 102028118-102028118 on Build GRCh38
rs370789247 Chr.7: 102028087-102028087 on Build GRCh38
rs755470058 Chr.7: 102028044-102028044 on Build GRCh38

View Full Product Details