Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129421554-129421781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB12
Entrez Gene ID
90288
Gene Name
EF-hand calcium binding domain 12
Gene Aliases
C3orf25
UniGene
Hs.652347
Gene Chromosome Location
Chr.3: 129401321-129428651 on Build GRCh38
Transcripts
NM_207307.2 XM_011513293.2 XM_011513294.2 XM_011513295.2 XM_011513297.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464942 c.135C>T p.F45F Chr.3: 129421718-129421718 on Build GRCh38
1038074 c.172C>T p.R58* Chr.3: 129421681-129421681 on Build GRCh38
86758 c.72C>T p.P24P Chr.3: 129421781-129421781 on Build GRCh38
6351296 c.197A>G p.E66G Chr.3: 129421656-129421656 on Build GRCh38
6651843 c.196G>A p.E66K Chr.3: 129421657-129421657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78778825 Chr.3: 129421683-129421683 on Build GRCh38
rs771372680 Chr.3: 129421621-129421621 on Build GRCh38
rs375275961 Chr.3: 129421702-129421702 on Build GRCh38
rs746231171 Chr.3: 129421741-129421741 on Build GRCh38
rs573087364 Chr.3: 129421747-129421747 on Build GRCh38
rs3774788 Chr.3: 129421646-129421646 on Build GRCh38
rs200277744 Chr.3: 129421615-129421615 on Build GRCh38
rs959794237 Chr.3: 129421663-129421663 on Build GRCh38
rs758388077 Chr.3: 129421655-129421655 on Build GRCh38
rs567796567 Chr.3: 129421647-129421647 on Build GRCh38
rs868483889 Chr.3: 129421748-129421748 on Build GRCh38
rs914599068 Chr.3: 129421577-129421577 on Build GRCh38
rs768550235 Chr.3: 129421660-129421660 on Build GRCh38
rs760320863 Chr.3: 129421686-129421686 on Build GRCh38
rs3774787 Chr.3: 129421656-129421656 on Build GRCh38
rs181124030 Chr.3: 129421662-129421662 on Build GRCh38
rs757734203 Chr.3: 129421730-129421730 on Build GRCh38
rs79535854 Chr.3: 129421701-129421701 on Build GRCh38
rs571623088 Chr.3: 129421680-129421680 on Build GRCh38
rs779428255 Chr.3: 129421576-129421576 on Build GRCh38
rs58932597 Chr.3: 129421746-129421746 on Build GRCh38
rs369063445 Chr.3: 129421745-129421745 on Build GRCh38
rs80203242 Chr.3: 129421704-129421704 on Build GRCh38
rs781429617 Chr.3: 129421630-129421630 on Build GRCh38
rs779000923 Chr.3: 129421618-129421618 on Build GRCh38
rs766196850 Chr.3: 129421693-129421693 on Build GRCh38
rs61752596 Chr.3: 129421681-129421681 on Build GRCh38
rs369186238 Chr.3: 129421644-129421644 on Build GRCh38
rs771027931 Chr.3: 129421598-129421598 on Build GRCh38
rs753416184 Chr.3: 129421588-129421588 on Build GRCh38
rs747302131 Chr.3: 129421620-129421620 on Build GRCh38
rs758964730 Chr.3: 129421612-129421612 on Build GRCh38
rs781551201 Chr.3: 129421736-129421736 on Build GRCh38
rs373478645 Chr.3: 129421684-129421684 on Build GRCh38

View Full Product Details