Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20959116-20959359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH3
Entrez Gene ID
55567
Gene Name
dynein axonemal heavy chain 3
Gene Aliases
DLP3, DNAHC3B, HEL-36, Hsadhc3, dnahc3-b
UniGene
Hs.526500
Gene Chromosome Location
Chr.16: 20933111-21159537 on Build GRCh38
Transcripts
NM_017539.2 XM_006721062.1 XM_011545882.2 XM_011545883.1 XM_011545885.2 XM_011545886.2 XM_011545888.2 XM_011545889.2 XM_017023426.1 XM_017023427.1 XM_017023428.1 XM_017023430.1 XM_017023432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1478604 c.10681A>G p.I3561V Chr.16: 20959324-20959324 on Build GRCh38
6564882 c.10826+8G>A p.? Chr.16: 20959171-20959171 on Build GRCh38
4627378 c.10680T>C p.P3560P Chr.16: 20959325-20959325 on Build GRCh38
1629811 c.10826+7T>C p.? Chr.16: 20959172-20959172 on Build GRCh38
6646627 c.10714G>T p.D3572Y Chr.16: 20959291-20959291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187774879 Chr.16: 20959174-20959174 on Build GRCh38
rs143956219 Chr.16: 20959289-20959289 on Build GRCh38
rs757652828 Chr.16: 20959155-20959155 on Build GRCh38
rs779755293 Chr.16: 20959276-20959276 on Build GRCh38
rs907177894 Chr.16: 20959300-20959300 on Build GRCh38
rs747998205 Chr.16: 20959197-20959197 on Build GRCh38
rs757844714 Chr.16: 20959270-20959270 on Build GRCh38
rs759520585 Chr.16: 20959207-20959207 on Build GRCh38
rs183710704 Chr.16: 20959201-20959201 on Build GRCh38
rs751000676 Chr.16: 20959264-20959264 on Build GRCh38
rs751238068 Chr.16: 20959280-20959280 on Build GRCh38
rs980346268 Chr.16: 20959161-20959161 on Build GRCh38
rs754318612 Chr.16: 20959151-20959151 on Build GRCh38
rs370491314 Chr.16: 20959135-20959135 on Build GRCh38
rs945857631 Chr.16: 20959263-20959269 on Build GRCh38
rs768797540 Chr.16: 20959337-20959337 on Build GRCh38
rs760935801 Chr.16: 20959252-20959252 on Build GRCh38
rs754629012 Chr.16: 20959288-20959288 on Build GRCh38
rs749286385 Chr.16: 20959302-20959302 on Build GRCh38
rs957007739 Chr.16: 20959238-20959238 on Build GRCh38
rs919437527 Chr.16: 20959209-20959209 on Build GRCh38
rs1045773820 Chr.16: 20959295-20959295 on Build GRCh38
rs764480676 Chr.16: 20959150-20959150 on Build GRCh38
rs866712566 Chr.16: 20959204-20959204 on Build GRCh38
rs767701712 Chr.16: 20959221-20959221 on Build GRCh38
rs368233531 Chr.16: 20959213-20959213 on Build GRCh38
rs765601438 Chr.16: 20959261-20959261 on Build GRCh38
rs760772025 Chr.16: 20959322-20959322 on Build GRCh38
rs775657876 Chr.16: 20959251-20959251 on Build GRCh38
rs772117284 Chr.16: 20959307-20959307 on Build GRCh38
rs191570218 Chr.16: 20959198-20959198 on Build GRCh38
rs868308321 Chr.16: 20959323-20959323 on Build GRCh38
rs754785450 Chr.16: 20959327-20959332 on Build GRCh38
rs776617532 Chr.16: 20959188-20959188 on Build GRCh38
rs190043471 Chr.16: 20959253-20959253 on Build GRCh38
rs753633198 Chr.16: 20959225-20959227 on Build GRCh38
rs746964153 Chr.16: 20959285-20959285 on Build GRCh38
rs768543696 Chr.16: 20959184-20959184 on Build GRCh38
rs34121503 Chr.16: 20959254-20959254 on Build GRCh38
rs746730307 Chr.16: 20959182-20959182 on Build GRCh38
rs747833121 Chr.16: 20959298-20959298 on Build GRCh38
rs745305717 Chr.16: 20959170-20959170 on Build GRCh38
rs150878924 Chr.16: 20959311-20959311 on Build GRCh38
rs751309838 Chr.16: 20959256-20959257 on Build GRCh38
rs777908315 Chr.16: 20959326-20959327 on Build GRCh38
rs769646154 Chr.16: 20959299-20959299 on Build GRCh38
rs757943247 Chr.16: 20959173-20959173 on Build GRCh38
rs779501797 Chr.16: 20959159-20959159 on Build GRCh38
rs757642945 Chr.16: 20959256-20959256 on Build GRCh38
rs140362873 Chr.16: 20959312-20959312 on Build GRCh38

View Full Product Details