Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36133131-36133378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
UniGene
Hs.606040
Gene Chromosome Location
Chr.18: 36129874-36177931 on Build GRCh38
Transcripts
NM_001242875.2 NM_001242876.2 NM_001242877.2 NM_001242878.2 NM_001242879.2 NM_001324465.1 NM_001324466.1 NM_001324467.1 NM_001324468.1 NM_018255.3 NR_040110.2 NR_136897.1 NR_136898.1 NR_137173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734851 c.197delG p.W66fs*1 Chr.18: 36133296-36133296 on Build GRCh38
7215513 c.217+8T>A p.? Chr.18: 36133324-36133324 on Build GRCh38
7415806 c.213T>A p.D71E Chr.18: 36133312-36133312 on Build GRCh38
4855039 c.174C>A p.T58T Chr.18: 36133273-36133273 on Build GRCh38
3734850 c.197delG p.W66fs*1 Chr.18: 36133296-36133296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765570747 Chr.18: 36133194-36133194 on Build GRCh38
rs146054918 Chr.18: 36133243-36133243 on Build GRCh38
rs898615980 Chr.18: 36133260-36133260 on Build GRCh38
rs753671102 Chr.18: 36133314-36133314 on Build GRCh38
rs979602144 Chr.18: 36133292-36133292 on Build GRCh38
rs747081813 Chr.18: 36133246-36133246 on Build GRCh38
rs749167262 Chr.18: 36133234-36133234 on Build GRCh38
rs779050765 Chr.18: 36133245-36133245 on Build GRCh38
rs202058551 Chr.18: 36133326-36133326 on Build GRCh38
rs770156126 Chr.18: 36133268-36133268 on Build GRCh38
rs150889303 Chr.18: 36133273-36133273 on Build GRCh38
rs779742209 Chr.18: 36133229-36133229 on Build GRCh38
rs139365470 Chr.18: 36133284-36133284 on Build GRCh38
rs184955852 Chr.18: 36133216-36133216 on Build GRCh38
rs1054756686 Chr.18: 36133168-36133168 on Build GRCh38
rs747548674 Chr.18: 36133244-36133246 on Build GRCh38
rs1785904 Chr.18: 36133169-36133169 on Build GRCh38
rs138888523 Chr.18: 36133244-36133244 on Build GRCh38
rs867996760 Chr.18: 36133247-36133247 on Build GRCh38
rs143686274 Chr.18: 36133171-36133171 on Build GRCh38
rs937218093 Chr.18: 36133302-36133302 on Build GRCh38
rs77729794 Chr.18: 36133222-36133222 on Build GRCh38
rs147222042 Chr.18: 36133179-36133179 on Build GRCh38
rs570228898 Chr.18: 36133278-36133278 on Build GRCh38
rs781163169 Chr.18: 36133331-36133331 on Build GRCh38
rs553697188 Chr.18: 36133277-36133277 on Build GRCh38
rs766954544 Chr.18: 36133282-36133282 on Build GRCh38
rs368406114 Chr.18: 36133309-36133309 on Build GRCh38
rs374884304 Chr.18: 36133344-36133344 on Build GRCh38
rs780895486 Chr.18: 36133213-36133213 on Build GRCh38
rs531678922 Chr.18: 36133195-36133195 on Build GRCh38
rs1048119802 Chr.18: 36133184-36133184 on Build GRCh38
rs1785905 Chr.18: 36133324-36133324 on Build GRCh38
rs763483033 Chr.18: 36133196-36133196 on Build GRCh38
rs368342716 Chr.18: 36133311-36133311 on Build GRCh38
rs778960872 Chr.18: 36133317-36133317 on Build GRCh38
rs763528269 Chr.18: 36133197-36133197 on Build GRCh38
rs763397524 Chr.18: 36133274-36133274 on Build GRCh38
rs776891738 Chr.18: 36133249-36133249 on Build GRCh38
rs773508864 Chr.18: 36133219-36133220 on Build GRCh38
rs757313096 Chr.18: 36133329-36133329 on Build GRCh38
rs141467947 Chr.18: 36133261-36133261 on Build GRCh38
rs1029577126 Chr.18: 36133182-36133182 on Build GRCh38
rs150024269 Chr.18: 36133307-36133307 on Build GRCh38
rs548070178 Chr.18: 36133201-36133201 on Build GRCh38
rs193081734 Chr.18: 36133208-36133208 on Build GRCh38

View Full Product Details