Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186613203-186613442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSG
Entrez Gene ID
197
Gene Name
alpha 2-HS glycoprotein
Gene Aliases
A2HS, AHS, FETUA, HSGA
UniGene
Hs.324746
Gene Chromosome Location
Chr.3: 186612928-186621318 on Build GRCh38
Transcripts
NM_001622.2 XM_017005840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042315 c.96C>T p.C32C Chr.3: 186613237-186613237 on Build GRCh38
6202541 c.206_207insG p.P70fs*10 Chr.3: 186613347-186613348 on Build GRCh38
7169313 c.144C>T p.I48I Chr.3: 186613285-186613285 on Build GRCh38
5688861 c.160T>A p.W54R Chr.3: 186613301-186613301 on Build GRCh38
6304987 c.180G>T p.L60F Chr.3: 186613321-186613321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776954754 Chr.3: 186613394-186613394 on Build GRCh38
rs771984910 Chr.3: 186613265-186613265 on Build GRCh38
rs962441625 Chr.3: 186613375-186613375 on Build GRCh38
rs777775803 Chr.3: 186613333-186613333 on Build GRCh38
rs987469127 Chr.3: 186613410-186613410 on Build GRCh38
rs780932666 Chr.3: 186613234-186613234 on Build GRCh38
rs986506968 Chr.3: 186613397-186613397 on Build GRCh38
rs745960136 Chr.3: 186613248-186613248 on Build GRCh38
rs965006114 Chr.3: 186613348-186613348 on Build GRCh38
rs1030440516 Chr.3: 186613364-186613364 on Build GRCh38
rs775843967 Chr.3: 186613269-186613269 on Build GRCh38
rs779762172 Chr.3: 186613229-186613229 on Build GRCh38
rs55962811 Chr.3: 186613422-186613422 on Build GRCh38
rs368781076 Chr.3: 186613367-186613367 on Build GRCh38
rs751252782 Chr.3: 186613308-186613308 on Build GRCh38
rs1004026351 Chr.3: 186613359-186613359 on Build GRCh38
rs141102051 Chr.3: 186613340-186613340 on Build GRCh38
rs769790254 Chr.3: 186613238-186613238 on Build GRCh38
rs777116283 Chr.3: 186613285-186613285 on Build GRCh38
rs1003867227 Chr.3: 186613338-186613338 on Build GRCh38
rs374708959 Chr.3: 186613343-186613343 on Build GRCh38
rs6774836 Chr.3: 186613378-186613378 on Build GRCh38
rs371743399 Chr.3: 186613330-186613330 on Build GRCh38
rs765801914 Chr.3: 186613306-186613306 on Build GRCh38
rs750828408 Chr.3: 186613302-186613302 on Build GRCh38
rs372710730 Chr.3: 186613399-186613399 on Build GRCh38
rs769025778 Chr.3: 186613388-186613388 on Build GRCh38
rs749060402 Chr.3: 186613237-186613237 on Build GRCh38
rs377000409 Chr.3: 186613230-186613230 on Build GRCh38
rs772980949 Chr.3: 186613244-186613244 on Build GRCh38
rs775683987 Chr.3: 186613350-186613350 on Build GRCh38
rs150486317 Chr.3: 186613349-186613349 on Build GRCh38
rs747965880 Chr.3: 186613325-186613325 on Build GRCh38
rs775522111 Chr.3: 186613274-186613274 on Build GRCh38
rs754601222 Chr.3: 186613317-186613317 on Build GRCh38
rs113040467 Chr.3: 186613355-186613355 on Build GRCh38
rs750484262 Chr.3: 186613272-186613272 on Build GRCh38
rs142527838 Chr.3: 186613231-186613231 on Build GRCh38
rs757807788 Chr.3: 186613226-186613226 on Build GRCh38
rs751378404 Chr.3: 186613339-186613339 on Build GRCh38
rs536966827 Chr.3: 186613405-186613405 on Build GRCh38
rs551743520 Chr.3: 186613295-186613295 on Build GRCh38
rs763034104 Chr.3: 186613275-186613275 on Build GRCh38
rs762220981 Chr.3: 186613287-186613287 on Build GRCh38
rs570498140 Chr.3: 186613305-186613305 on Build GRCh38
rs199602010 Chr.3: 186613395-186613395 on Build GRCh38

View Full Product Details