Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40190717-40190990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPGEFL1
Entrez Gene ID
51195
Gene Name
Rap guanine nucleotide exchange factor like 1
Gene Aliases
Link-GEFII
UniGene
Hs.632254
Gene Chromosome Location
Chr.17: 40177010-40195656 on Build GRCh38
Transcripts
NM_001303533.1 NM_001303534.1 NM_016339.4 XM_006721932.3 XM_006721935.3 XM_006721937.3 XM_006721938.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146641 c.712C>T p.H238Y Chr.17: 40190757-40190757 on Build GRCh38
6762918 c.704G>A p.R235Q Chr.17: 40190749-40190749 on Build GRCh38
5473885 c.685T>G p.F229V Chr.17: 40190730-40190730 on Build GRCh38
978849 c.703C>T p.R235* Chr.17: 40190748-40190748 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901943259 Chr.17: 40190933-40190933 on Build GRCh38
rs768184580 Chr.17: 40190903-40190903 on Build GRCh38
rs1006497642 Chr.17: 40190830-40190830 on Build GRCh38
rs749198202 Chr.17: 40190772-40190772 on Build GRCh38
rs572833215 Chr.17: 40190824-40190824 on Build GRCh38
rs759678149 Chr.17: 40190807-40190807 on Build GRCh38
rs557476181 Chr.17: 40190749-40190749 on Build GRCh38
rs750231375 Chr.17: 40190868-40190868 on Build GRCh38
rs773294247 Chr.17: 40190769-40190769 on Build GRCh38
rs763081388 Chr.17: 40190924-40190925 on Build GRCh38
rs555660696 Chr.17: 40190839-40190839 on Build GRCh38
rs562318070 Chr.17: 40190922-40190922 on Build GRCh38
rs780891353 Chr.17: 40190744-40190744 on Build GRCh38
rs924499759 Chr.17: 40190921-40190921 on Build GRCh38
rs919054413 Chr.17: 40190856-40190856 on Build GRCh38
rs947021286 Chr.17: 40190909-40190909 on Build GRCh38
rs972202972 Chr.17: 40190843-40190843 on Build GRCh38
rs776851003 Chr.17: 40190794-40190794 on Build GRCh38
rs768739826 Chr.17: 40190777-40190777 on Build GRCh38
rs573585776 Chr.17: 40190871-40190871 on Build GRCh38
rs760781943 Chr.17: 40190782-40190782 on Build GRCh38
rs762668758 Chr.17: 40190915-40190915 on Build GRCh38
rs765082937 Chr.17: 40190858-40190858 on Build GRCh38
rs774483392 Chr.17: 40190778-40190778 on Build GRCh38
rs1016513153 Chr.17: 40190831-40190831 on Build GRCh38
rs879099523 Chr.17: 40190764-40190765 on Build GRCh38
rs199682021 Chr.17: 40190739-40190739 on Build GRCh38
rs533549270 Chr.17: 40190833-40190833 on Build GRCh38
rs1028958684 Chr.17: 40190822-40190822 on Build GRCh38
rs766439773 Chr.17: 40190790-40190790 on Build GRCh38
rs1045062153 Chr.17: 40190925-40190925 on Build GRCh38
rs959785611 Chr.17: 40190862-40190862 on Build GRCh38
rs548239903 Chr.17: 40190809-40190809 on Build GRCh38
rs753079370 Chr.17: 40190812-40190812 on Build GRCh38
rs985794933 Chr.17: 40190784-40190784 on Build GRCh38
rs377128108 Chr.17: 40190919-40190920 on Build GRCh38
rs144655242 Chr.17: 40190896-40190896 on Build GRCh38
rs775430226 Chr.17: 40190767-40190767 on Build GRCh38

View Full Product Details