Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112647282-112647529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP7
Entrez Gene ID
51574
Gene Name
La ribonucleoprotein domain family member 7
Gene Aliases
ALAZS, HDCMA18P, PIP7S
UniGene
Hs.713663
Gene Chromosome Location
Chr.4: 112636964-112657592 on Build GRCh38
Transcripts
NM_001267039.1 NM_015454.2 NM_016648.3 NR_049768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1256334 c.794C>A p.S265Y Chr.4: 112647346-112647346 on Build GRCh38
5974962 c.743T>G p.I248S Chr.4: 112647295-112647295 on Build GRCh38
5474669 c.813G>T p.Q271H Chr.4: 112647365-112647365 on Build GRCh38
4450919 c.922delC p.R309fs*4 Chr.4: 112647474-112647474 on Build GRCh38
7111156 c.799G>A p.E267K Chr.4: 112647351-112647351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753043822 Chr.4: 112647371-112647371 on Build GRCh38
rs1023594873 Chr.4: 112647406-112647406 on Build GRCh38
rs770950781 Chr.4: 112647423-112647423 on Build GRCh38
rs969327433 Chr.4: 112647445-112647445 on Build GRCh38
rs371035787 Chr.4: 112647478-112647478 on Build GRCh38
rs373167325 Chr.4: 112647322-112647322 on Build GRCh38
rs375829533 Chr.4: 112647366-112647366 on Build GRCh38
rs900139242 Chr.4: 112647349-112647349 on Build GRCh38
rs754655165 Chr.4: 112647491-112647491 on Build GRCh38
rs751017218 Chr.4: 112647486-112647486 on Build GRCh38
rs373219872 Chr.4: 112647323-112647323 on Build GRCh38
rs201250325 Chr.4: 112647387-112647387 on Build GRCh38
rs763010665 Chr.4: 112647326-112647326 on Build GRCh38
rs764744775 Chr.4: 112647471-112647471 on Build GRCh38
rs771193812 Chr.4: 112647334-112647334 on Build GRCh38
rs756166445 Chr.4: 112647371-112647376 on Build GRCh38
rs764693485 Chr.4: 112647380-112647380 on Build GRCh38
rs745604512 Chr.4: 112647391-112647392 on Build GRCh38
rs774437783 Chr.4: 112647424-112647424 on Build GRCh38
rs757874580 Chr.4: 112647480-112647499 on Build GRCh38
rs990748843 Chr.4: 112647470-112647470 on Build GRCh38
rs754365322 Chr.4: 112647473-112647473 on Build GRCh38
rs765820560 Chr.4: 112647481-112647481 on Build GRCh38
rs373634124 Chr.4: 112647401-112647401 on Build GRCh38
rs1027546928 Chr.4: 112647346-112647346 on Build GRCh38
rs763929099 Chr.4: 112647380-112647381 on Build GRCh38
rs866288154 Chr.4: 112647477-112647477 on Build GRCh38
rs761264490 Chr.4: 112647374-112647374 on Build GRCh38
rs541057628 Chr.4: 112647460-112647460 on Build GRCh38
rs937970407 Chr.4: 112647442-112647442 on Build GRCh38
rs62317770 Chr.4: 112647388-112647388 on Build GRCh38
rs772265784 Chr.4: 112647436-112647436 on Build GRCh38
rs775781419 Chr.4: 112647448-112647448 on Build GRCh38
rs747933441 Chr.4: 112647391-112647391 on Build GRCh38
rs778420137 Chr.4: 112647386-112647387 on Build GRCh38
rs536947693 Chr.4: 112647319-112647319 on Build GRCh38
rs780993494 Chr.4: 112647317-112647317 on Build GRCh38
rs529243678 Chr.4: 112647367-112647367 on Build GRCh38
rs767188889 Chr.4: 112647493-112647493 on Build GRCh38
rs754602989 Chr.4: 112647389-112647389 on Build GRCh38
rs767792487 Chr.4: 112647369-112647369 on Build GRCh38
rs773773118 Chr.4: 112647362-112647362 on Build GRCh38
rs749413494 Chr.4: 112647408-112647408 on Build GRCh38
rs774408363 Chr.4: 112647342-112647342 on Build GRCh38
rs201212143 Chr.4: 112647378-112647378 on Build GRCh38
rs757958464 Chr.4: 112647385-112647385 on Build GRCh38
rs997074760 Chr.4: 112647350-112647350 on Build GRCh38
rs577197566 Chr.4: 112647426-112647426 on Build GRCh38
rs979666067 Chr.4: 112647472-112647472 on Build GRCh38
rs201727777 Chr.4: 112647402-112647402 on Build GRCh38
rs562023320 Chr.4: 112647347-112647347 on Build GRCh38

View Full Product Details