Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6970514-6970755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHB2
Entrez Gene ID
11331
Gene Name
prohibitin 2
Gene Aliases
BAP, BCAP37, Bap37, PNAS-141, REA, hBAP, p22
UniGene
Hs.504620
Gene Chromosome Location
Chr.12: 6965352-6970823 on Build GRCh38
Transcripts
NM_001144831.1 NM_001267700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744825 c.26C>T p.A9V Chr.12: 6970518-6970518 on Build GRCh38
6901799 c.7C>T p.Q3* Chr.12: 6970537-6970537 on Build GRCh38
3464721 c.9G>A p.Q3Q Chr.12: 6970535-6970535 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370707865 Chr.12: 6970554-6970554 on Build GRCh38
rs11541468 Chr.12: 6970664-6970664 on Build GRCh38
rs782457762 Chr.12: 6970605-6970605 on Build GRCh38
rs782489596 Chr.12: 6970614-6970614 on Build GRCh38
rs782224419 Chr.12: 6970700-6970700 on Build GRCh38
rs782477805 Chr.12: 6970733-6970733 on Build GRCh38
rs782484300 Chr.12: 6970681-6970681 on Build GRCh38
rs782425183 Chr.12: 6970581-6970581 on Build GRCh38
rs937076158 Chr.12: 6970644-6970644 on Build GRCh38
rs375302364 Chr.12: 6970572-6970572 on Build GRCh38
rs958279876 Chr.12: 6970567-6970567 on Build GRCh38
rs1009851213 Chr.12: 6970703-6970703 on Build GRCh38
rs982698993 Chr.12: 6970736-6970736 on Build GRCh38
rs971289636 Chr.12: 6970582-6970582 on Build GRCh38
rs781960116 Chr.12: 6970570-6970570 on Build GRCh38
rs78952305 Chr.12: 6970654-6970654 on Build GRCh38
rs782768325 Chr.12: 6970589-6970589 on Build GRCh38
rs781814872 Chr.12: 6970675-6970675 on Build GRCh38
rs35084368 Chr.12: 6970699-6970700 on Build GRCh38
rs782268104 Chr.12: 6970540-6970540 on Build GRCh38
rs782180993 Chr.12: 6970670-6970670 on Build GRCh38
rs376534055 Chr.12: 6970553-6970553 on Build GRCh38
rs782109515 Chr.12: 6970660-6970660 on Build GRCh38
rs782094225 Chr.12: 6970587-6970588 on Build GRCh38
rs782014932 Chr.12: 6970593-6970593 on Build GRCh38
rs782719021 Chr.12: 6970584-6970584 on Build GRCh38
rs1021199666 Chr.12: 6970717-6970717 on Build GRCh38
rs781976262 Chr.12: 6970585-6970585 on Build GRCh38
rs782574509 Chr.12: 6970679-6970679 on Build GRCh38
rs1032384013 Chr.12: 6970696-6970696 on Build GRCh38
rs11541466 Chr.12: 6970587-6970587 on Build GRCh38
rs782792427 Chr.12: 6970653-6970653 on Build GRCh38
rs782633394 Chr.12: 6970722-6970722 on Build GRCh38
rs193061358 Chr.12: 6970623-6970623 on Build GRCh38
rs782524743 Chr.12: 6970656-6970656 on Build GRCh38
rs1055394225 Chr.12: 6970649-6970649 on Build GRCh38

View Full Product Details