Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94827569-94827830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1102665 c.478C>T p.P160S Chr.8: 94827753-94827753 on Build GRCh38
4595717 c.479C>T p.P160L Chr.8: 94827754-94827754 on Build GRCh38
5875146 c.497C>T p.P166L Chr.8: 94827772-94827772 on Build GRCh38
5450496 c.498C>T p.P166P Chr.8: 94827773-94827773 on Build GRCh38
6871717 c.515G>A p.S172N Chr.8: 94827790-94827790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35707070 Chr.8: 94827800-94827801 on Build GRCh38
rs765163153 Chr.8: 94827748-94827748 on Build GRCh38
rs758426501 Chr.8: 94827774-94827774 on Build GRCh38
rs781019304 Chr.8: 94827786-94827786 on Build GRCh38
rs368541220 Chr.8: 94827726-94827726 on Build GRCh38
rs774736396 Chr.8: 94827707-94827707 on Build GRCh38
rs775894659 Chr.8: 94827723-94827723 on Build GRCh38
rs750330835 Chr.8: 94827773-94827773 on Build GRCh38
rs796052157 Chr.8: 94827759-94827759 on Build GRCh38
rs112990773 Chr.8: 94827628-94827628 on Build GRCh38
rs923690253 Chr.8: 94827664-94827664 on Build GRCh38
rs1024652410 Chr.8: 94827607-94827607 on Build GRCh38
rs954982287 Chr.8: 94827747-94827747 on Build GRCh38
rs1032448290 Chr.8: 94827600-94827600 on Build GRCh38
rs771425594 Chr.8: 94827695-94827695 on Build GRCh38
rs747698703 Chr.8: 94827689-94827689 on Build GRCh38
rs772324188 Chr.8: 94827717-94827717 on Build GRCh38
rs978258948 Chr.8: 94827662-94827662 on Build GRCh38
rs372034182 Chr.8: 94827745-94827745 on Build GRCh38
rs766817593 Chr.8: 94827710-94827714 on Build GRCh38
rs368018334 Chr.8: 94827667-94827667 on Build GRCh38
rs766071672 Chr.8: 94827776-94827776 on Build GRCh38
rs780822843 Chr.8: 94827673-94827673 on Build GRCh38
rs746664328 Chr.8: 94827617-94827620 on Build GRCh38
rs376730024 Chr.8: 94827716-94827716 on Build GRCh38
rs761205918 Chr.8: 94827679-94827682 on Build GRCh38
rs542936494 Chr.8: 94827641-94827641 on Build GRCh38
rs942658130 Chr.8: 94827767-94827767 on Build GRCh38
rs754643472 Chr.8: 94827777-94827777 on Build GRCh38
rs761904801 Chr.8: 94827746-94827746 on Build GRCh38
rs987704042 Chr.8: 94827802-94827802 on Build GRCh38
rs761516772 Chr.8: 94827640-94827640 on Build GRCh38

View Full Product Details