Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72601868-72602139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKA1
Entrez Gene ID
5255
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Gene Aliases
PHKA
UniGene
Hs.201379
Gene Chromosome Location
Chr.X: 72578814-72714181 on Build GRCh38
Transcripts
NM_001122670.1 NM_001172436.1 NM_002637.3 XM_006724661.2

SNP Detail

SNP ID Chromosome Location
rs782387067 Chr.X: 72602038-72602038 on Build GRCh38
rs782566958 Chr.X: 72602002-72602002 on Build GRCh38
rs370105009 Chr.X: 72601926-72601926 on Build GRCh38
rs1037583701 Chr.X: 72601980-72601980 on Build GRCh38
rs782107667 Chr.X: 72602036-72602036 on Build GRCh38
rs782638867 Chr.X: 72601950-72601950 on Build GRCh38
rs1047174037 Chr.X: 72602075-72602075 on Build GRCh38
rs199530028 Chr.X: 72602035-72602035 on Build GRCh38
rs137936397 Chr.X: 72602025-72602025 on Build GRCh38
rs782344973 Chr.X: 72601995-72601995 on Build GRCh38
rs782267710 Chr.X: 72601970-72601970 on Build GRCh38
rs782191011 Chr.X: 72602020-72602020 on Build GRCh38
rs782361223 Chr.X: 72601895-72601895 on Build GRCh38
rs184195764 Chr.X: 72601901-72601901 on Build GRCh38
rs929986098 Chr.X: 72602031-72602031 on Build GRCh38
rs781947907 Chr.X: 72602064-72602064 on Build GRCh38
rs186704637 Chr.X: 72601920-72601920 on Build GRCh38

View Full Product Details