Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49809060-49809289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT7
Entrez Gene ID
11143
Gene Name
lysine acetyltransferase 7
Gene Aliases
HBO1, HBOA, MYST2, ZC2HC7
UniGene
Hs.21907
Gene Chromosome Location
Chr.17: 49788619-49829096 on Build GRCh38
Transcripts
NM_001199155.1 NM_001199156.1 NM_001199157.1 NM_001199158.1 NM_007067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795755 c.666G>A p.V222V Chr.17: 49809121-49809121 on Build GRCh38
707003 c.664-1G>T p.? Chr.17: 49809118-49809118 on Build GRCh38
5755068 c.670G>A p.A224T Chr.17: 49809125-49809125 on Build GRCh38
1610432 c.710C>T p.S237F Chr.17: 49809165-49809165 on Build GRCh38
7294095 c.712C>T p.H238Y Chr.17: 49809167-49809167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182075628 Chr.17: 49809252-49809252 on Build GRCh38
rs377599635 Chr.17: 49809167-49809167 on Build GRCh38
rs753817770 Chr.17: 49809246-49809246 on Build GRCh38
rs777255663 Chr.17: 49809192-49809192 on Build GRCh38
rs112316627 Chr.17: 49809238-49809238 on Build GRCh38
rs754705481 Chr.17: 49809097-49809097 on Build GRCh38
rs370241626 Chr.17: 49809199-49809199 on Build GRCh38
rs752582459 Chr.17: 49809101-49809101 on Build GRCh38
rs769035683 Chr.17: 49809187-49809187 on Build GRCh38
rs747438558 Chr.17: 49809178-49809178 on Build GRCh38
rs779152890 Chr.17: 49809253-49809253 on Build GRCh38
rs763878666 Chr.17: 49809245-49809245 on Build GRCh38
rs773955785 Chr.17: 49809212-49809212 on Build GRCh38
rs375058247 Chr.17: 49809156-49809156 on Build GRCh38
rs770483324 Chr.17: 49809204-49809204 on Build GRCh38
rs760586779 Chr.17: 49809241-49809241 on Build GRCh38
rs757244076 Chr.17: 49809247-49809247 on Build GRCh38
rs777905284 Chr.17: 49809180-49809182 on Build GRCh38
rs767206335 Chr.17: 49809234-49809234 on Build GRCh38
rs535118323 Chr.17: 49809102-49809102 on Build GRCh38
rs547575711 Chr.17: 49809163-49809163 on Build GRCh38
rs985045886 Chr.17: 49809233-49809233 on Build GRCh38
rs141200367 Chr.17: 49809146-49809146 on Build GRCh38
rs777808325 Chr.17: 49809109-49809109 on Build GRCh38
rs1035683546 Chr.17: 49809181-49809181 on Build GRCh38
rs767332852 Chr.17: 49809099-49809099 on Build GRCh38
rs759210470 Chr.17: 49809223-49809223 on Build GRCh38
rs749313607 Chr.17: 49809134-49809134 on Build GRCh38
rs956296843 Chr.17: 49809185-49809185 on Build GRCh38
rs779210077 Chr.17: 49809151-49809151 on Build GRCh38

View Full Product Details