Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1980104-1980329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOXO1
Entrez Gene ID
124056
Gene Name
NADPH oxidase organizer 1
Gene Aliases
P41NOX, P41NOXA, P41NOXB, P41NOXC, SH3PXD5, SNX28
UniGene
Hs.191762
Gene Chromosome Location
Chr.16: 1978917-1982751 on Build GRCh38
Transcripts
NM_001267721.1 NM_144603.3 NM_172167.2 NM_172168.2 XM_005255099.4 XM_017022927.1 XM_017022928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376329 c.468_469delCA p.H156fs*21 Chr.16: 1980114-1980115 on Build GRCh38
3754761 c.457C>T p.L153F Chr.16: 1980126-1980126 on Build GRCh38
4128770 c.445G>A p.A149T Chr.16: 1980138-1980138 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953442115 Chr.16: 1980306-1980311 on Build GRCh38
rs201337633 Chr.16: 1980202-1980202 on Build GRCh38
rs201282851 Chr.16: 1980204-1980204 on Build GRCh38
rs774586915 Chr.16: 1980196-1980196 on Build GRCh38
rs1013527949 Chr.16: 1980235-1980235 on Build GRCh38
rs1011668454 Chr.16: 1980298-1980298 on Build GRCh38
rs879519186 Chr.16: 1980151-1980151 on Build GRCh38
rs976463944 Chr.16: 1980143-1980143 on Build GRCh38
rs1021264877 Chr.16: 1980230-1980230 on Build GRCh38
rs769420511 Chr.16: 1980303-1980304 on Build GRCh38
rs963839231 Chr.16: 1980215-1980215 on Build GRCh38
rs528002653 Chr.16: 1980303-1980303 on Build GRCh38
rs375214922 Chr.16: 1980124-1980124 on Build GRCh38
rs758153013 Chr.16: 1980141-1980141 on Build GRCh38
rs1008102961 Chr.16: 1980211-1980211 on Build GRCh38
rs530197455 Chr.16: 1980179-1980179 on Build GRCh38
rs898390107 Chr.16: 1980157-1980157 on Build GRCh38
rs760784545 Chr.16: 1980214-1980214 on Build GRCh38
rs553734924 Chr.16: 1980286-1980286 on Build GRCh38
rs111267512 Chr.16: 1980158-1980158 on Build GRCh38
rs61729118 Chr.16: 1980126-1980126 on Build GRCh38
rs1011160058 Chr.16: 1980224-1980224 on Build GRCh38
rs148232934 Chr.16: 1980260-1980260 on Build GRCh38
rs369956618 Chr.16: 1980231-1980231 on Build GRCh38
rs527577865 Chr.16: 1980265-1980265 on Build GRCh38
rs531378694 Chr.16: 1980234-1980234 on Build GRCh38
rs759395937 Chr.16: 1980225-1980225 on Build GRCh38
rs552247687 Chr.16: 1980296-1980297 on Build GRCh38
rs777368177 Chr.16: 1980156-1980156 on Build GRCh38
rs367786229 Chr.16: 1980164-1980164 on Build GRCh38
rs915941323 Chr.16: 1980302-1980302 on Build GRCh38
rs991679611 Chr.16: 1980291-1980291 on Build GRCh38
rs766225026 Chr.16: 1980221-1980221 on Build GRCh38
rs747768713 Chr.16: 1980133-1980133 on Build GRCh38
rs776128511 Chr.16: 1980171-1980171 on Build GRCh38
rs759971058 Chr.16: 1980122-1980122 on Build GRCh38
rs994096218 Chr.16: 1980186-1980186 on Build GRCh38
rs1020425398 Chr.16: 1980300-1980300 on Build GRCh38
rs770389719 Chr.16: 1980169-1980169 on Build GRCh38
rs778418100 Chr.16: 1980127-1980127 on Build GRCh38
rs1050563771 Chr.16: 1980185-1980185 on Build GRCh38
rs769010540 Chr.16: 1980189-1980189 on Build GRCh38
rs551088000 Chr.16: 1980249-1980249 on Build GRCh38
rs753872817 Chr.16: 1980223-1980223 on Build GRCh38
rs761986635 Chr.16: 1980201-1980201 on Build GRCh38
rs928927122 Chr.16: 1980279-1980279 on Build GRCh38

View Full Product Details