Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 78602411-78602666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU4F1
Entrez Gene ID
5457
Gene Name
POU class 4 homeobox 1
Gene Aliases
BRN3A, Oct-T1, RDC-1, brn-3A
UniGene
Hs.654522
Gene Chromosome Location
Chr.13: 78599092-78603560 on Build GRCh38
Transcripts
NM_006237.3 NR_047001.1

Gene Symbol
RNF219-AS1
Entrez Gene ID
100874222
Gene Name
RNF219 antisense RNA 1
Gene Aliases
POU4F1-AS1
UniGene
Hs.493062
Gene Chromosome Location
Chr.13: 78054855-78617325 on Build GRCh38
Transcripts
NM_006237.3 NR_047001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6852509 c.251C>T p.T84M Chr.13: 78602424-78602424 on Build GRCh38
5823429 c.198C>A p.I66I Chr.13: 78602477-78602477 on Build GRCh38
3469841 c.224C>T p.P75L Chr.13: 78602451-78602451 on Build GRCh38
4800777 c.124-2A>T p.? Chr.13: 78602553-78602553 on Build GRCh38
6320362 c.179C>A p.A60E Chr.13: 78602496-78602496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764267688 Chr.13: 78602502-78602502 on Build GRCh38
rs561881656 Chr.13: 78602640-78602640 on Build GRCh38
rs1023819809 Chr.13: 78602505-78602505 on Build GRCh38
rs1027118318 Chr.13: 78602572-78602572 on Build GRCh38
rs1028611003 Chr.13: 78602610-78602610 on Build GRCh38
rs999560525 Chr.13: 78602476-78602476 on Build GRCh38
rs756049643 Chr.13: 78602555-78602555 on Build GRCh38
rs190922162 Chr.13: 78602612-78602612 on Build GRCh38
rs868371322 Chr.13: 78602487-78602487 on Build GRCh38
rs370292716 Chr.13: 78602596-78602596 on Build GRCh38
rs751771213 Chr.13: 78602517-78602517 on Build GRCh38
rs202049972 Chr.13: 78602537-78602537 on Build GRCh38
rs752859017 Chr.13: 78602495-78602495 on Build GRCh38
rs749531923 Chr.13: 78602575-78602575 on Build GRCh38
rs200546833 Chr.13: 78602598-78602598 on Build GRCh38
rs780951016 Chr.13: 78602549-78602549 on Build GRCh38
rs142455133 Chr.13: 78602539-78602539 on Build GRCh38
rs745603488 Chr.13: 78602554-78602554 on Build GRCh38
rs758452608 Chr.13: 78602498-78602498 on Build GRCh38
rs760987880 Chr.13: 78602436-78602436 on Build GRCh38
rs538156451 Chr.13: 78602484-78602484 on Build GRCh38
rs372182236 Chr.13: 78602597-78602597 on Build GRCh38
rs147611646 Chr.13: 78602625-78602625 on Build GRCh38
rs558380820 Chr.13: 78602489-78602489 on Build GRCh38
rs547701923 Chr.13: 78602556-78602556 on Build GRCh38
rs1037160220 Chr.13: 78602588-78602588 on Build GRCh38
rs899722751 Chr.13: 78602541-78602541 on Build GRCh38
rs201126908 Chr.13: 78602474-78602474 on Build GRCh38
rs768182522 Chr.13: 78602581-78602581 on Build GRCh38
rs779926376 Chr.13: 78602559-78602559 on Build GRCh38
rs1020110294 Chr.13: 78602628-78602628 on Build GRCh38
rs74097352 Chr.13: 78602483-78602483 on Build GRCh38
rs748846937 Chr.13: 78602579-78602579 on Build GRCh38
rs766763681 Chr.13: 78602438-78602438 on Build GRCh38
rs376644798 Chr.13: 78602452-78602452 on Build GRCh38
rs73537043 Chr.13: 78602635-78602635 on Build GRCh38

View Full Product Details