Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37937160-37937429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT1L1
Entrez Gene ID
137362
Gene Name
glutamic-oxaloacetic transaminase 1-like 1
Gene Aliases
-
UniGene
Hs.380740
Gene Chromosome Location
Chr.8: 37933548-37940502 on Build GRCh38
Transcripts
NM_152413.2 XM_005273399.3 XM_006716285.3 XM_011544391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3648800 c.418G>A p.G140R Chr.8: 37937378-37937378 on Build GRCh38
3648798 c.495C>T p.P165P Chr.8: 37937301-37937301 on Build GRCh38
5678626 c.446C>A p.T149K Chr.8: 37937350-37937350 on Build GRCh38
4753058 c.470A>G p.D157G Chr.8: 37937326-37937326 on Build GRCh38
4856454 c.497A>G p.D166G Chr.8: 37937299-37937299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966543055 Chr.8: 37937239-37937239 on Build GRCh38
rs758018572 Chr.8: 37937400-37937401 on Build GRCh38
rs752487987 Chr.8: 37937329-37937332 on Build GRCh38
rs770606644 Chr.8: 37937270-37937270 on Build GRCh38
rs533801133 Chr.8: 37937264-37937264 on Build GRCh38
rs568888757 Chr.8: 37937263-37937263 on Build GRCh38
rs370723407 Chr.8: 37937279-37937279 on Build GRCh38
rs868168203 Chr.8: 37937377-37937377 on Build GRCh38
rs974741340 Chr.8: 37937248-37937248 on Build GRCh38
rs189485549 Chr.8: 37937184-37937184 on Build GRCh38
rs772444398 Chr.8: 37937355-37937355 on Build GRCh38
rs775730379 Chr.8: 37937281-37937281 on Build GRCh38
rs557845117 Chr.8: 37937297-37937297 on Build GRCh38
rs747300324 Chr.8: 37937368-37937368 on Build GRCh38
rs981011405 Chr.8: 37937271-37937271 on Build GRCh38
rs529582480 Chr.8: 37937211-37937211 on Build GRCh38
rs374016210 Chr.8: 37937301-37937301 on Build GRCh38
rs752720932 Chr.8: 37937398-37937398 on Build GRCh38
rs756786046 Chr.8: 37937342-37937342 on Build GRCh38
rs1018849612 Chr.8: 37937350-37937350 on Build GRCh38
rs533748703 Chr.8: 37937373-37937373 on Build GRCh38
rs181030975 Chr.8: 37937251-37937251 on Build GRCh38
rs926966473 Chr.8: 37937379-37937379 on Build GRCh38
rs953902960 Chr.8: 37937259-37937259 on Build GRCh38
rs368566290 Chr.8: 37937372-37937372 on Build GRCh38
rs764957626 Chr.8: 37937279-37937281 on Build GRCh38
rs765236057 Chr.8: 37937382-37937382 on Build GRCh38
rs765319837 Chr.8: 37937300-37937300 on Build GRCh38
rs374467683 Chr.8: 37937353-37937353 on Build GRCh38
rs566597559 Chr.8: 37937307-37937307 on Build GRCh38
rs760149854 Chr.8: 37937275-37937275 on Build GRCh38
rs767352858 Chr.8: 37937396-37937396 on Build GRCh38
rs756004933 Chr.8: 37937320-37937320 on Build GRCh38
rs202201309 Chr.8: 37937327-37937327 on Build GRCh38
rs776128178 Chr.8: 37937356-37937356 on Build GRCh38
rs367552836 Chr.8: 37937306-37937306 on Build GRCh38
rs769348490 Chr.8: 37937252-37937252 on Build GRCh38
rs756982078 Chr.8: 37937406-37937406 on Build GRCh38
rs61735752 Chr.8: 37937384-37937384 on Build GRCh38
rs764004832 Chr.8: 37937401-37937401 on Build GRCh38
rs774978036 Chr.8: 37937273-37937273 on Build GRCh38
rs539578990 Chr.8: 37937282-37937282 on Build GRCh38
rs748999268 Chr.8: 37937329-37937329 on Build GRCh38
rs932962444 Chr.8: 37937312-37937312 on Build GRCh38
rs778568424 Chr.8: 37937351-37937351 on Build GRCh38
rs761857388 Chr.8: 37937299-37937299 on Build GRCh38
rs753473533 Chr.8: 37937404-37937404 on Build GRCh38
rs774701711 Chr.8: 37937335-37937335 on Build GRCh38
rs764214355 Chr.8: 37937292-37937292 on Build GRCh38
rs773349377 Chr.8: 37937389-37937389 on Build GRCh38
rs368386251 Chr.8: 37937399-37937399 on Build GRCh38
rs888842509 Chr.8: 37937334-37937334 on Build GRCh38
rs936517690 Chr.8: 37937280-37937280 on Build GRCh38
rs535388951 Chr.8: 37937319-37937319 on Build GRCh38

View Full Product Details