Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247815116-247815359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR14A16
Entrez Gene ID
284532
Gene Name
olfactory receptor family 14 subfamily A member 16
Gene Aliases
OR5AT1
UniGene
Hs.626620
Gene Chromosome Location
Chr.1: 247814800-247815729 on Build GRCh38
Transcripts
NM_001001966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3966439 c.379C>A p.P127T Chr.1: 247815351-247815351 on Build GRCh38
4030610 c.533A>T p.D178V Chr.1: 247815197-247815197 on Build GRCh38
5036453 c.521A>C p.Q174P Chr.1: 247815209-247815209 on Build GRCh38
6062886 c.595C>T p.L199F Chr.1: 247815135-247815135 on Build GRCh38
3486804 c.403G>A p.D135N Chr.1: 247815327-247815327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745386234 Chr.1: 247815283-247815283 on Build GRCh38
rs771054740 Chr.1: 247815204-247815204 on Build GRCh38
rs749856984 Chr.1: 247815310-247815310 on Build GRCh38
rs745910297 Chr.1: 247815321-247815321 on Build GRCh38
rs771359251 Chr.1: 247815316-247815316 on Build GRCh38
rs746728262 Chr.1: 247815165-247815165 on Build GRCh38
rs201678616 Chr.1: 247815305-247815305 on Build GRCh38
rs754698306 Chr.1: 247815272-247815272 on Build GRCh38
rs748492840 Chr.1: 247815194-247815194 on Build GRCh38
rs375198428 Chr.1: 247815242-247815244 on Build GRCh38
rs780852701 Chr.1: 247815279-247815279 on Build GRCh38
rs200138400 Chr.1: 247815241-247815241 on Build GRCh38
rs74152818 Chr.1: 247815215-247815215 on Build GRCh38
rs75017395 Chr.1: 247815284-247815284 on Build GRCh38
rs767730748 Chr.1: 247815230-247815230 on Build GRCh38
rs764923003 Chr.1: 247815248-247815248 on Build GRCh38
rs750448350 Chr.1: 247815252-247815252 on Build GRCh38
rs751052236 Chr.1: 247815268-247815268 on Build GRCh38
rs749067685 Chr.1: 247815239-247815241 on Build GRCh38
rs1046097850 Chr.1: 247815256-247815256 on Build GRCh38
rs770231134 Chr.1: 247815195-247815195 on Build GRCh38
rs781421072 Chr.1: 247815179-247815179 on Build GRCh38
rs753607782 Chr.1: 247815233-247815233 on Build GRCh38
rs35979231 Chr.1: 247815240-247815242 on Build GRCh38
rs141705060 Chr.1: 247815329-247815329 on Build GRCh38
rs867096409 Chr.1: 247815198-247815198 on Build GRCh38
rs774437577 Chr.1: 247815317-247815317 on Build GRCh38
rs570397976 Chr.1: 247815267-247815267 on Build GRCh38
rs763427823 Chr.1: 247815202-247815202 on Build GRCh38
rs773445245 Chr.1: 247815239-247815240 on Build GRCh38
rs769407268 Chr.1: 247815294-247815295 on Build GRCh38
rs777986261 Chr.1: 247815309-247815309 on Build GRCh38
rs768354012 Chr.1: 247815177-247815177 on Build GRCh38
rs201817284 Chr.1: 247815260-247815260 on Build GRCh38
rs759768896 Chr.1: 247815227-247815227 on Build GRCh38
rs113112592 Chr.1: 247815242-247815242 on Build GRCh38
rs761709000 Chr.1: 247815240-247815240 on Build GRCh38

View Full Product Details