Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49681829-49682072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT1
Entrez Gene ID
3276
Gene Name
protein arginine methyltransferase 1
Gene Aliases
ANM1, HCP1, HRMT1L2, IR1B4
UniGene
Hs.20521
Gene Chromosome Location
Chr.19: 49676166-49688450 on Build GRCh38
Transcripts
NM_001207042.2 NM_001536.5 NM_198318.4 NR_033397.4 XM_005258842.1 XM_017026734.1 XM_017026735.1 XM_017026736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4285362 c.328G>A p.G110R Chr.19: 49682045-49682045 on Build GRCh38
4482287 c.186C>T p.L62L Chr.19: 49681975-49681975 on Build GRCh38
1395305 c.243T>C p.F81F Chr.19: 49682032-49682032 on Build GRCh38
7126304 c.198G>T p.M66I Chr.19: 49681915-49681915 on Build GRCh38
2753081 c.143G>A p.R48H Chr.19: 49681932-49681932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056829207 Chr.19: 49682047-49682047 on Build GRCh38
rs1804486 Chr.19: 49681980-49681980 on Build GRCh38
rs777703888 Chr.19: 49681887-49681887 on Build GRCh38
rs763480376 Chr.19: 49681870-49681870 on Build GRCh38
rs746776701 Chr.19: 49681934-49681943 on Build GRCh38
rs775323970 Chr.19: 49681968-49681968 on Build GRCh38
rs763717229 Chr.19: 49681984-49681984 on Build GRCh38
rs749876886 Chr.19: 49681894-49681894 on Build GRCh38
rs377108642 Chr.19: 49681886-49681886 on Build GRCh38
rs766142574 Chr.19: 49682020-49682020 on Build GRCh38
rs370547880 Chr.19: 49681905-49681905 on Build GRCh38
rs373144744 Chr.19: 49681885-49681885 on Build GRCh38
rs753023530 Chr.19: 49682036-49682036 on Build GRCh38
rs143736498 Chr.19: 49681882-49681882 on Build GRCh38
rs905248991 Chr.19: 49682015-49682015 on Build GRCh38
rs886889677 Chr.19: 49681849-49681849 on Build GRCh38
rs554576824 Chr.19: 49681969-49681969 on Build GRCh38
rs746258178 Chr.19: 49681924-49681924 on Build GRCh38
rs760264054 Chr.19: 49681867-49681867 on Build GRCh38
rs777594676 Chr.19: 49681882-49681889 on Build GRCh38
rs539807277 Chr.19: 49682044-49682044 on Build GRCh38
rs758217712 Chr.19: 49682008-49682008 on Build GRCh38
rs61732965 Chr.19: 49682052-49682052 on Build GRCh38
rs781412610 Chr.19: 49681883-49681883 on Build GRCh38
rs1047197455 Chr.19: 49681848-49681848 on Build GRCh38
rs750220873 Chr.19: 49681878-49681878 on Build GRCh38
rs753860642 Chr.19: 49681874-49681874 on Build GRCh38
rs759914608 Chr.19: 49681860-49681860 on Build GRCh38
rs199719552 Chr.19: 49681999-49681999 on Build GRCh38
rs904818817 Chr.19: 49682014-49682014 on Build GRCh38
rs868411595 Chr.19: 49682016-49682016 on Build GRCh38
rs143467686 Chr.19: 49681942-49681942 on Build GRCh38
rs750309006 Chr.19: 49682007-49682007 on Build GRCh38
rs200342298 Chr.19: 49681895-49681895 on Build GRCh38
rs757128021 Chr.19: 49681875-49681875 on Build GRCh38
rs765168424 Chr.19: 49682002-49682002 on Build GRCh38
rs775235752 Chr.19: 49681866-49681866 on Build GRCh38

View Full Product Details