Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 233016830-233017055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNX2
Entrez Gene ID
80003
Gene Name
pecanex homolog 2 (Drosophila)
Gene Aliases
PCNXL2
UniGene
Hs.370605
Gene Chromosome Location
Chr.1: 232984109-233295735 on Build GRCh38
Transcripts
NM_014801.3 XM_011544278.2 XM_017002391.1 XM_017002392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485283 c.4815A>G p.Q1605Q Chr.1: 233016945-233016945 on Build GRCh38
5796984 c.4801C>A p.L1601I Chr.1: 233016959-233016959 on Build GRCh38
4549593 c.4778G>A p.R1593Q Chr.1: 233016982-233016982 on Build GRCh38
3369611 c.4739T>A p.I1580N Chr.1: 233017021-233017021 on Build GRCh38
6841237 c.4777C>T p.R1593* Chr.1: 233016983-233016983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748423427 Chr.1: 233016906-233016906 on Build GRCh38
rs767665261 Chr.1: 233017001-233017001 on Build GRCh38
rs866390344 Chr.1: 233016891-233016891 on Build GRCh38
rs937067606 Chr.1: 233017025-233017025 on Build GRCh38
rs534347022 Chr.1: 233016957-233016957 on Build GRCh38
rs373400160 Chr.1: 233017013-233017013 on Build GRCh38
rs751751691 Chr.1: 233016881-233016881 on Build GRCh38
rs746406640 Chr.1: 233017016-233017016 on Build GRCh38
rs766206998 Chr.1: 233016968-233016968 on Build GRCh38
rs369652650 Chr.1: 233016934-233016934 on Build GRCh38
rs775967243 Chr.1: 233016953-233016953 on Build GRCh38
rs551811471 Chr.1: 233016926-233016926 on Build GRCh38
rs538249519 Chr.1: 233017021-233017021 on Build GRCh38
rs537352943 Chr.1: 233016987-233016987 on Build GRCh38
rs375561904 Chr.1: 233016901-233016901 on Build GRCh38
rs1007009686 Chr.1: 233016912-233016912 on Build GRCh38
rs992049785 Chr.1: 233016958-233016958 on Build GRCh38
rs866800257 Chr.1: 233016991-233016991 on Build GRCh38
rs768177831 Chr.1: 233016949-233016949 on Build GRCh38
rs780123571 Chr.1: 233016877-233016877 on Build GRCh38
rs761569730 Chr.1: 233016956-233016956 on Build GRCh38
rs756408220 Chr.1: 233017002-233017002 on Build GRCh38
rs575594777 Chr.1: 233017008-233017008 on Build GRCh38
rs188866535 Chr.1: 233016935-233016935 on Build GRCh38
rs776256928 Chr.1: 233017022-233017022 on Build GRCh38
rs781072655 Chr.1: 233016905-233016905 on Build GRCh38
rs774843785 Chr.1: 233016938-233016938 on Build GRCh38
rs923444995 Chr.1: 233017024-233017024 on Build GRCh38
rs749446491 Chr.1: 233017004-233017004 on Build GRCh38
rs1033415664 Chr.1: 233016898-233016898 on Build GRCh38
rs759980974 Chr.1: 233016941-233016941 on Build GRCh38
rs751554663 Chr.1: 233016983-233016983 on Build GRCh38
rs747602738 Chr.1: 233017023-233017023 on Build GRCh38
rs927102187 Chr.1: 233017005-233017005 on Build GRCh38
rs567156613 Chr.1: 233016988-233016988 on Build GRCh38
rs769865641 Chr.1: 233016918-233016918 on Build GRCh38
rs200950369 Chr.1: 233016959-233016959 on Build GRCh38
rs369127953 Chr.1: 233016978-233016978 on Build GRCh38
rs556044650 Chr.1: 233017007-233017007 on Build GRCh38
rs536105916 Chr.1: 233017003-233017003 on Build GRCh38

View Full Product Details