Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70435196-70435449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NODAL
Entrez Gene ID
4838
Gene Name
nodal growth differentiation factor
Gene Aliases
HTX5
UniGene
Hs.370414
Gene Chromosome Location
Chr.10: 70431936-70441709 on Build GRCh38
Transcripts
NM_018055.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2160423 c.834C>T p.G278G Chr.10: 70435343-70435343 on Build GRCh38
2160422 c.859G>C p.E287Q Chr.10: 70435318-70435318 on Build GRCh38
7196710 c.823C>G p.R275G Chr.10: 70435354-70435354 on Build GRCh38
5520893 c.742C>T p.R248W Chr.10: 70435435-70435435 on Build GRCh38
32311 c.835G>A p.E279K Chr.10: 70435342-70435342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375311498 Chr.10: 70435267-70435267 on Build GRCh38
rs150892417 Chr.10: 70435313-70435313 on Build GRCh38
rs760173416 Chr.10: 70435273-70435273 on Build GRCh38
rs763525961 Chr.10: 70435307-70435307 on Build GRCh38
rs369869149 Chr.10: 70435271-70435271 on Build GRCh38
rs774058854 Chr.10: 70435361-70435361 on Build GRCh38
rs886831582 Chr.10: 70435260-70435260 on Build GRCh38
rs878855044 Chr.10: 70435285-70435285 on Build GRCh38
rs944753811 Chr.10: 70435301-70435301 on Build GRCh38
rs781366461 Chr.10: 70435354-70435354 on Build GRCh38
rs2231958 Chr.10: 70435266-70435266 on Build GRCh38
rs755116310 Chr.10: 70435342-70435342 on Build GRCh38
rs747091068 Chr.10: 70435244-70435244 on Build GRCh38
rs771688576 Chr.10: 70435376-70435376 on Build GRCh38
rs121909283 Chr.10: 70435399-70435399 on Build GRCh38
rs775212204 Chr.10: 70435384-70435384 on Build GRCh38
rs773894278 Chr.10: 70435233-70435233 on Build GRCh38
rs187711450 Chr.10: 70435242-70435242 on Build GRCh38
rs769914386 Chr.10: 70435253-70435253 on Build GRCh38
rs773281767 Chr.10: 70435261-70435261 on Build GRCh38
rs770958126 Chr.10: 70435360-70435360 on Build GRCh38
rs375503197 Chr.10: 70435330-70435330 on Build GRCh38
rs193203393 Chr.10: 70435341-70435341 on Build GRCh38
rs780042490 Chr.10: 70435272-70435272 on Build GRCh38
rs757713122 Chr.10: 70435308-70435308 on Build GRCh38
rs1046746392 Chr.10: 70435258-70435258 on Build GRCh38
rs749265748 Chr.10: 70435358-70435358 on Build GRCh38
rs780038392 Chr.10: 70435243-70435243 on Build GRCh38
rs555563029 Chr.10: 70435353-70435353 on Build GRCh38
rs747666961 Chr.10: 70435266-70435272 on Build GRCh38
rs139409082 Chr.10: 70435400-70435400 on Build GRCh38
rs758598737 Chr.10: 70435326-70435326 on Build GRCh38
rs370796035 Chr.10: 70435309-70435309 on Build GRCh38
rs760261207 Chr.10: 70435392-70435392 on Build GRCh38
rs745659788 Chr.10: 70435367-70435367 on Build GRCh38

View Full Product Details