Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15563411-15563672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HACL1
Entrez Gene ID
26061
Gene Name
2-hydroxyacyl-CoA lyase 1
Gene Aliases
2-HPCL, HPCL, HPCL2, PHYH2
UniGene
Hs.63290
Gene Chromosome Location
Chr.3: 15560704-15601852 on Build GRCh38
Transcripts
NM_001284413.1 NM_001284415.1 NM_001284416.1 NM_012260.3 NR_104315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5398506 c.1623C>T p.S541S Chr.3: 15563439-15563439 on Build GRCh38
4444893 c.1604C>T p.P535L Chr.3: 15563458-15563458 on Build GRCh38
1040337 c.1534T>G p.L512V Chr.3: 15563528-15563528 on Build GRCh38
4114776 c.1602A>G p.T534T Chr.3: 15563460-15563460 on Build GRCh38
4114775 c.1636C>G p.L546V Chr.3: 15563426-15563426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372266033 Chr.3: 15563532-15563532 on Build GRCh38
rs774488253 Chr.3: 15563628-15563628 on Build GRCh38
rs574249207 Chr.3: 15563496-15563496 on Build GRCh38
rs75831568 Chr.3: 15563507-15563507 on Build GRCh38
rs373765837 Chr.3: 15563454-15563454 on Build GRCh38
rs772746261 Chr.3: 15563442-15563442 on Build GRCh38
rs762732269 Chr.3: 15563445-15563445 on Build GRCh38
rs555074049 Chr.3: 15563508-15563510 on Build GRCh38
rs754840690 Chr.3: 15563534-15563534 on Build GRCh38
rs999656083 Chr.3: 15563521-15563521 on Build GRCh38
rs777383596 Chr.3: 15563589-15563589 on Build GRCh38
rs186020952 Chr.3: 15563448-15563448 on Build GRCh38
rs766118795 Chr.3: 15563589-15563589 on Build GRCh38
rs891137481 Chr.3: 15563625-15563625 on Build GRCh38
rs890223686 Chr.3: 15563516-15563516 on Build GRCh38
rs764799164 Chr.3: 15563438-15563438 on Build GRCh38
rs752768853 Chr.3: 15563493-15563493 on Build GRCh38
rs532040608 Chr.3: 15563610-15563610 on Build GRCh38
rs772724245 Chr.3: 15563442-15563442 on Build GRCh38
rs760080495 Chr.3: 15563456-15563456 on Build GRCh38
rs769180870 Chr.3: 15563569-15563569 on Build GRCh38
rs1038059939 Chr.3: 15563490-15563490 on Build GRCh38
rs751408852 Chr.3: 15563594-15563595 on Build GRCh38
rs748140011 Chr.3: 15563556-15563556 on Build GRCh38
rs751383477 Chr.3: 15563533-15563533 on Build GRCh38
rs368735834 Chr.3: 15563515-15563515 on Build GRCh38
rs893486987 Chr.3: 15563597-15563597 on Build GRCh38
rs1052393608 Chr.3: 15563618-15563618 on Build GRCh38
rs916720212 Chr.3: 15563590-15563590 on Build GRCh38
rs768143327 Chr.3: 15563476-15563476 on Build GRCh38
rs1043803360 Chr.3: 15563601-15563601 on Build GRCh38
rs781118545 Chr.3: 15563555-15563555 on Build GRCh38
rs753978123 Chr.3: 15563513-15563513 on Build GRCh38
rs115046099 Chr.3: 15563609-15563609 on Build GRCh38
rs370345174 Chr.3: 15563440-15563440 on Build GRCh38
rs982294069 Chr.3: 15563623-15563623 on Build GRCh38
rs776487772 Chr.3: 15563458-15563458 on Build GRCh38
rs969595810 Chr.3: 15563619-15563619 on Build GRCh38
rs769607941 Chr.3: 15563541-15563541 on Build GRCh38

View Full Product Details