Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55637408-55637671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10P1
Entrez Gene ID
121130
Gene Name
olfactory receptor family 10 subfamily P member 1
Gene Aliases
OR10P1P, OR10P2P, OR10P3P, OR12-7, OST701
UniGene
Hs.554551
Gene Chromosome Location
Chr.12: 55636892-55637833 on Build GRCh38
Transcripts
NM_206899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147270 c.561G>T p.R187S Chr.12: 55637452-55637452 on Build GRCh38
4147269 c.560G>A p.R187K Chr.12: 55637451-55637451 on Build GRCh38
6025322 c.647T>A p.V216D Chr.12: 55637538-55637538 on Build GRCh38
4147268 c.559A>G p.R187G Chr.12: 55637450-55637450 on Build GRCh38
84169 c.663C>T p.R221R Chr.12: 55637554-55637554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753878405 Chr.12: 55637595-55637595 on Build GRCh38
rs746076508 Chr.12: 55637521-55637521 on Build GRCh38
rs77750365 Chr.12: 55637554-55637554 on Build GRCh38
rs771970961 Chr.12: 55637610-55637610 on Build GRCh38
rs780175736 Chr.12: 55637527-55637527 on Build GRCh38
rs7970885 Chr.12: 55637489-55637489 on Build GRCh38
rs747889943 Chr.12: 55637549-55637549 on Build GRCh38
rs374333859 Chr.12: 55637475-55637475 on Build GRCh38
rs141032808 Chr.12: 55637480-55637480 on Build GRCh38
rs1046028478 Chr.12: 55637567-55637567 on Build GRCh38
rs765883468 Chr.12: 55637483-55637483 on Build GRCh38
rs374844386 Chr.12: 55637622-55637622 on Build GRCh38
rs762402694 Chr.12: 55637635-55637635 on Build GRCh38
rs954065500 Chr.12: 55637649-55637649 on Build GRCh38
rs756328996 Chr.12: 55637524-55637524 on Build GRCh38
rs528637565 Chr.12: 55637573-55637573 on Build GRCh38
rs926070325 Chr.12: 55637503-55637503 on Build GRCh38
rs772940707 Chr.12: 55637479-55637479 on Build GRCh38
rs781218612 Chr.12: 55637505-55637505 on Build GRCh38
rs749215030 Chr.12: 55637455-55637455 on Build GRCh38
rs776178000 Chr.12: 55637618-55637618 on Build GRCh38
rs768642018 Chr.12: 55637470-55637470 on Build GRCh38
rs769402040 Chr.12: 55637623-55637623 on Build GRCh38
rs757496236 Chr.12: 55637428-55637428 on Build GRCh38
rs752131085 Chr.12: 55637574-55637574 on Build GRCh38
rs140326016 Chr.12: 55637598-55637598 on Build GRCh38
rs759120415 Chr.12: 55637556-55637556 on Build GRCh38
rs371380402 Chr.12: 55637594-55637594 on Build GRCh38
rs750735614 Chr.12: 55637641-55637641 on Build GRCh38
rs546484739 Chr.12: 55637552-55637552 on Build GRCh38
rs757884989 Chr.12: 55637585-55637585 on Build GRCh38
rs562248481 Chr.12: 55637613-55637613 on Build GRCh38
rs745892411 Chr.12: 55637440-55637440 on Build GRCh38
rs770548804 Chr.12: 55637617-55637617 on Build GRCh38
rs775176418 Chr.12: 55637633-55637633 on Build GRCh38
rs774308493 Chr.12: 55637537-55637537 on Build GRCh38
rs764709944 Chr.12: 55637491-55637491 on Build GRCh38
rs768517537 Chr.12: 55637535-55637535 on Build GRCh38
rs377460542 Chr.12: 55637553-55637553 on Build GRCh38
rs750817849 Chr.12: 55637593-55637593 on Build GRCh38
rs767120434 Chr.12: 55637478-55637478 on Build GRCh38
rs199780580 Chr.12: 55637638-55637638 on Build GRCh38
rs76940436 Chr.12: 55637534-55637534 on Build GRCh38
rs751906346 Chr.12: 55637499-55637499 on Build GRCh38
rs779246737 Chr.12: 55637532-55637535 on Build GRCh38
rs755076681 Chr.12: 55637597-55637597 on Build GRCh38
rs761605993 Chr.12: 55637476-55637476 on Build GRCh38
rs146703207 Chr.12: 55637586-55637586 on Build GRCh38
rs769935432 Chr.12: 55637452-55637452 on Build GRCh38
rs748047790 Chr.12: 55637603-55637603 on Build GRCh38
rs1004506907 Chr.12: 55637566-55637566 on Build GRCh38
rs200538833 Chr.12: 55637550-55637550 on Build GRCh38
rs753358058 Chr.12: 55637488-55637488 on Build GRCh38
rs757727573 Chr.12: 55637501-55637501 on Build GRCh38
rs781568825 Chr.12: 55637433-55637433 on Build GRCh38

View Full Product Details