Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 53824442-53824689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2B
Entrez Gene ID
84643
Gene Name
kinesin family member 2B
Gene Aliases
-
UniGene
Hs.226805
Gene Chromosome Location
Chr.17: 53822878-53825212 on Build GRCh38
Transcripts
NM_032559.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3193865 c.1526G>A p.G509D Chr.17: 53824559-53824559 on Build GRCh38
4067920 c.1534T>A p.S512T Chr.17: 53824567-53824567 on Build GRCh38
3889982 c.1450G>A p.A484T Chr.17: 53824483-53824483 on Build GRCh38
4528822 c.1566G>A p.P522P Chr.17: 53824599-53824599 on Build GRCh38
295336 c.1620delA p.K542fs*2 Chr.17: 53824653-53824653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776724226 Chr.17: 53824578-53824578 on Build GRCh38
rs912018606 Chr.17: 53824509-53824509 on Build GRCh38
rs139231634 Chr.17: 53824497-53824497 on Build GRCh38
rs1054773121 Chr.17: 53824642-53824642 on Build GRCh38
rs374320401 Chr.17: 53824657-53824657 on Build GRCh38
rs745698463 Chr.17: 53824549-53824549 on Build GRCh38
rs766858144 Chr.17: 53824522-53824522 on Build GRCh38
rs143245314 Chr.17: 53824544-53824544 on Build GRCh38
rs763960000 Chr.17: 53824554-53824554 on Build GRCh38
rs777650812 Chr.17: 53824658-53824658 on Build GRCh38
rs150898451 Chr.17: 53824573-53824573 on Build GRCh38
rs772927955 Chr.17: 53824625-53824625 on Build GRCh38
rs775566987 Chr.17: 53824476-53824476 on Build GRCh38
rs773474575 Chr.17: 53824586-53824586 on Build GRCh38
rs898835619 Chr.17: 53824664-53824664 on Build GRCh38
rs753437090 Chr.17: 53824616-53824616 on Build GRCh38
rs370994388 Chr.17: 53824471-53824471 on Build GRCh38
rs138313576 Chr.17: 53824593-53824593 on Build GRCh38
rs755466034 Chr.17: 53824539-53824539 on Build GRCh38
rs149373861 Chr.17: 53824617-53824617 on Build GRCh38
rs752110971 Chr.17: 53824528-53824528 on Build GRCh38
rs754949894 Chr.17: 53824649-53824649 on Build GRCh38
rs764855772 Chr.17: 53824628-53824628 on Build GRCh38
rs1046161794 Chr.17: 53824634-53824634 on Build GRCh38
rs374451533 Chr.17: 53824508-53824508 on Build GRCh38
rs751318038 Chr.17: 53824648-53824648 on Build GRCh38
rs775476052 Chr.17: 53824469-53824469 on Build GRCh38
rs567762338 Chr.17: 53824579-53824579 on Build GRCh38
rs945268597 Chr.17: 53824613-53824613 on Build GRCh38
rs768094492 Chr.17: 53824604-53824604 on Build GRCh38
rs149644944 Chr.17: 53824641-53824641 on Build GRCh38
rs372345633 Chr.17: 53824637-53824637 on Build GRCh38
rs186833308 Chr.17: 53824540-53824540 on Build GRCh38
rs745728329 Chr.17: 53824652-53824653 on Build GRCh38
rs774794184 Chr.17: 53824598-53824598 on Build GRCh38
rs374095045 Chr.17: 53824659-53824659 on Build GRCh38
rs768533876 Chr.17: 53824576-53824576 on Build GRCh38
rs774243494 Chr.17: 53824559-53824559 on Build GRCh38
rs778609949 Chr.17: 53824547-53824548 on Build GRCh38
rs757000396 Chr.17: 53824543-53824543 on Build GRCh38
rs553176548 Chr.17: 53824599-53824599 on Build GRCh38
rs769961143 Chr.17: 53824581-53824581 on Build GRCh38
rs768727956 Chr.17: 53824485-53824485 on Build GRCh38
rs773492584 Chr.17: 53824507-53824507 on Build GRCh38
rs762047480 Chr.17: 53824501-53824501 on Build GRCh38
rs765461122 Chr.17: 53824502-53824502 on Build GRCh38

View Full Product Details