Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108487866-108488117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA8
Entrez Gene ID
50640
Gene Name
patatin like phospholipase domain containing 8
Gene Aliases
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma
UniGene
Hs.617340
Gene Chromosome Location
Chr.7: 108470417-108528161 on Build GRCh38
Transcripts
NM_001256007.2 NM_001256008.2 NM_001256009.2 NM_001256010.2 NM_001256011.2 NM_015723.4 XM_005250396.4 XM_011516274.2 XM_011516275.2 XM_017012261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4472068 c.1759C>T p.P587S Chr.7: 108487878-108487878 on Build GRCh38
3631896 c.1700C>T p.T567I Chr.7: 108487937-108487937 on Build GRCh38
6751580 c.1734T>C p.F578F Chr.7: 108487903-108487903 on Build GRCh38
6751578 c.1722A>G p.T574T Chr.7: 108487915-108487915 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779916994 Chr.7: 108487985-108487985 on Build GRCh38
rs774128364 Chr.7: 108488004-108488004 on Build GRCh38
rs565461120 Chr.7: 108488008-108488008 on Build GRCh38
rs745456604 Chr.7: 108487978-108487978 on Build GRCh38
rs753285330 Chr.7: 108487955-108487955 on Build GRCh38
rs756351922 Chr.7: 108487934-108487934 on Build GRCh38
rs1000014467 Chr.7: 108488010-108488010 on Build GRCh38
rs768548603 Chr.7: 108488002-108488002 on Build GRCh38
rs754208244 Chr.7: 108487947-108487947 on Build GRCh38
rs769572138 Chr.7: 108487979-108487979 on Build GRCh38
rs761779041 Chr.7: 108487902-108487902 on Build GRCh38
rs1046816046 Chr.7: 108487973-108487973 on Build GRCh38
rs1031162252 Chr.7: 108488010-108488011 on Build GRCh38
rs766672877 Chr.7: 108487935-108487935 on Build GRCh38
rs749084206 Chr.7: 108487999-108487999 on Build GRCh38
rs774407467 Chr.7: 108487896-108487896 on Build GRCh38
rs1029018310 Chr.7: 108487945-108487945 on Build GRCh38
rs538281682 Chr.7: 108487924-108487924 on Build GRCh38
rs559611795 Chr.7: 108488034-108488034 on Build GRCh38
rs779264581 Chr.7: 108487953-108487953 on Build GRCh38
rs551722953 Chr.7: 108487959-108487959 on Build GRCh38
rs911941406 Chr.7: 108488072-108488072 on Build GRCh38
rs780772699 Chr.7: 108487968-108487968 on Build GRCh38
rs750608746 Chr.7: 108487925-108487925 on Build GRCh38
rs184013871 Chr.7: 108488066-108488066 on Build GRCh38
rs953855743 Chr.7: 108488073-108488073 on Build GRCh38
rs755439764 Chr.7: 108487951-108487951 on Build GRCh38
rs893941340 Chr.7: 108488029-108488029 on Build GRCh38

View Full Product Details