Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2568835-2569104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCE
Entrez Gene ID
23288
Gene Name
IQ motif containing E
Gene Aliases
1700028P05Rik
UniGene
Hs.520627
Gene Chromosome Location
Chr.7: 2558972-2614734 on Build GRCh38
Transcripts
NM_001287499.1 NM_001287500.1 NM_001287501.1 NM_001287502.1 NM_152558.4 XM_006715676.2 XM_011515242.1 XM_011515243.1 XM_017011902.1 XM_017011903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6177274 c.96G>T p.R32S Chr.7: 2568965-2568965 on Build GRCh38
6177273 c.125C>T p.S42L Chr.7: 2568994-2568994 on Build GRCh38
4846198 c.96G>A p.R32R Chr.7: 2568965-2568965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771630447 Chr.7: 2568927-2568927 on Build GRCh38
rs777307398 Chr.7: 2568941-2568944 on Build GRCh38
rs896249523 Chr.7: 2568891-2568891 on Build GRCh38
rs766363405 Chr.7: 2568983-2568983 on Build GRCh38
rs774312827 Chr.7: 2568863-2568863 on Build GRCh38
rs759988518 Chr.7: 2568947-2568947 on Build GRCh38
rs373608047 Chr.7: 2569031-2569031 on Build GRCh38
rs1013262733 Chr.7: 2568893-2568893 on Build GRCh38
rs761878243 Chr.7: 2568980-2568980 on Build GRCh38
rs372444042 Chr.7: 2568998-2568998 on Build GRCh38
rs376875364 Chr.7: 2569021-2569021 on Build GRCh38
rs765052494 Chr.7: 2569048-2569048 on Build GRCh38
rs201117529 Chr.7: 2568906-2568906 on Build GRCh38
rs746485113 Chr.7: 2568910-2568910 on Build GRCh38
rs761896390 Chr.7: 2569032-2569032 on Build GRCh38
rs546669369 Chr.7: 2569006-2569006 on Build GRCh38
rs369323781 Chr.7: 2569023-2569023 on Build GRCh38
rs369630946 Chr.7: 2569003-2569003 on Build GRCh38
rs763190190 Chr.7: 2568960-2568960 on Build GRCh38
rs938804023 Chr.7: 2568978-2568978 on Build GRCh38
rs751703419 Chr.7: 2568967-2568967 on Build GRCh38
rs566575196 Chr.7: 2569010-2569010 on Build GRCh38
rs775609832 Chr.7: 2568949-2568949 on Build GRCh38
rs1016140073 Chr.7: 2568864-2568864 on Build GRCh38
rs200840589 Chr.7: 2569000-2569000 on Build GRCh38
rs372239080 Chr.7: 2569011-2569011 on Build GRCh38
rs753718686 Chr.7: 2568986-2568986 on Build GRCh38
rs190076769 Chr.7: 2569081-2569081 on Build GRCh38
rs764203982 Chr.7: 2568963-2568963 on Build GRCh38
rs200043602 Chr.7: 2569034-2569034 on Build GRCh38
rs770213756 Chr.7: 2568948-2568948 on Build GRCh38
rs771627025 Chr.7: 2569036-2569036 on Build GRCh38
rs115130019 Chr.7: 2569050-2569050 on Build GRCh38
rs759517421 Chr.7: 2569037-2569037 on Build GRCh38
rs776581226 Chr.7: 2569035-2569035 on Build GRCh38
rs770504454 Chr.7: 2569005-2569005 on Build GRCh38
rs367632329 Chr.7: 2568995-2568995 on Build GRCh38
rs903543800 Chr.7: 2568861-2568861 on Build GRCh38
rs935212479 Chr.7: 2569067-2569067 on Build GRCh38
rs746645479 Chr.7: 2569029-2569030 on Build GRCh38
rs369909640 Chr.7: 2568902-2568902 on Build GRCh38
rs374023484 Chr.7: 2568877-2568877 on Build GRCh38
rs199648730 Chr.7: 2568994-2568994 on Build GRCh38
rs754772955 Chr.7: 2568992-2568992 on Build GRCh38
rs373052635 Chr.7: 2569024-2569024 on Build GRCh38
rs766307916 Chr.7: 2569016-2569016 on Build GRCh38
rs777261327 Chr.7: 2568938-2568938 on Build GRCh38
rs367938810 Chr.7: 2568901-2568901 on Build GRCh38
rs555132443 Chr.7: 2568898-2568898 on Build GRCh38

View Full Product Details