Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 124646893-124647166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF608
Entrez Gene ID
57507
Gene Name
zinc finger protein 608
Gene Aliases
NY-REN-36
UniGene
Hs.266616
Gene Chromosome Location
Chr.5: 124636913-124748824 on Build GRCh38
Transcripts
NM_020747.2 XM_005272037.2 XM_005272038.1 XM_005272039.2 XM_005272040.3 XM_011543520.2 XM_011543522.2 XM_011543523.2 XM_017009674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1432725 c.3465A>G p.K1155K Chr.5: 124646919-124646919 on Build GRCh38
3849747 c.3479C>T p.P1160L Chr.5: 124646905-124646905 on Build GRCh38
5975321 c.3249T>C p.Y1083Y Chr.5: 124647135-124647135 on Build GRCh38
1166906 c.3472T>A p.S1158T Chr.5: 124646912-124646912 on Build GRCh38
1060172 c.3412C>A p.L1138M Chr.5: 124646972-124646972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200127477 Chr.5: 124647006-124647006 on Build GRCh38
rs759979865 Chr.5: 124647045-124647045 on Build GRCh38
rs189382479 Chr.5: 124646934-124646934 on Build GRCh38
rs780782486 Chr.5: 124646980-124646980 on Build GRCh38
rs142977399 Chr.5: 124647011-124647011 on Build GRCh38
rs761882925 Chr.5: 124647091-124647091 on Build GRCh38
rs775950140 Chr.5: 124647060-124647060 on Build GRCh38
rs750144991 Chr.5: 124647099-124647099 on Build GRCh38
rs569222523 Chr.5: 124647118-124647118 on Build GRCh38
rs145806100 Chr.5: 124646990-124646990 on Build GRCh38
rs748930733 Chr.5: 124646981-124646981 on Build GRCh38
rs768268162 Chr.5: 124646993-124646993 on Build GRCh38
rs771772913 Chr.5: 124647023-124647023 on Build GRCh38
rs150431508 Chr.5: 124646968-124646968 on Build GRCh38
rs146130870 Chr.5: 124647081-124647081 on Build GRCh38
rs780053794 Chr.5: 124647110-124647110 on Build GRCh38
rs777331817 Chr.5: 124646941-124646941 on Build GRCh38
rs764151706 Chr.5: 124647078-124647078 on Build GRCh38
rs551955365 Chr.5: 124646979-124646979 on Build GRCh38
rs374453730 Chr.5: 124647054-124647054 on Build GRCh38
rs767801852 Chr.5: 124647092-124647092 on Build GRCh38
rs371545787 Chr.5: 124647044-124647044 on Build GRCh38
rs778510856 Chr.5: 124647119-124647119 on Build GRCh38
rs892187993 Chr.5: 124647085-124647085 on Build GRCh38
rs755956432 Chr.5: 124646983-124646983 on Build GRCh38
rs762985854 Chr.5: 124646939-124646939 on Build GRCh38
rs771197992 Chr.5: 124646914-124646914 on Build GRCh38
rs747719675 Chr.5: 124647123-124647123 on Build GRCh38
rs747737637 Chr.5: 124647016-124647016 on Build GRCh38
rs752737759 Chr.5: 124646936-124646936 on Build GRCh38
rs138950020 Chr.5: 124647082-124647082 on Build GRCh38
rs374429838 Chr.5: 124646935-124646935 on Build GRCh38
rs763505514 Chr.5: 124647063-124647063 on Build GRCh38
rs750244492 Chr.5: 124646982-124646982 on Build GRCh38
rs757950895 Chr.5: 124647126-124647126 on Build GRCh38
rs138281065 Chr.5: 124646992-124646992 on Build GRCh38
rs200980889 Chr.5: 124646987-124646987 on Build GRCh38

View Full Product Details