Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150374524-150374797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCOF1
Entrez Gene ID
6949
Gene Name
treacle ribosome biogenesis factor 1
Gene Aliases
MFD1, TCS, TCS1, treacle
UniGene
Hs.519672
Gene Chromosome Location
Chr.5: 150357639-150400308 on Build GRCh38
Transcripts
NM_000356.3 NM_001008657.2 NM_001135243.1 NM_001135244.1 NM_001135245.1 NM_001195141.1 XM_005268502.3 XM_005268503.3 XM_005268504.3 XM_005268505.3 XM_005268506.3 XM_005268507.3 XM_005268508.3 XM_005268509.3 XM_011537678.2 XM_017009792.1 XM_017009793.1 XM_017009794.1 XM_017009795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4345702 c.1196C>T p.A399V Chr.5: 150374729-150374729 on Build GRCh38
1696570 c.940C>T p.P314S Chr.5: 150374704-150374704 on Build GRCh38
6208793 c.1155A>G p.G385G Chr.5: 150374688-150374688 on Build GRCh38
6259044 c.1149G>T p.R383S Chr.5: 150374682-150374682 on Build GRCh38
3669428 c.1164A>T p.P388P Chr.5: 150374697-150374697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781642190 Chr.5: 150374590-150374590 on Build GRCh38
rs773945106 Chr.5: 150374699-150374699 on Build GRCh38
rs749626536 Chr.5: 150374576-150374576 on Build GRCh38
rs140204483 Chr.5: 150374750-150374750 on Build GRCh38
rs773266881 Chr.5: 150374694-150374694 on Build GRCh38
rs764117099 Chr.5: 150374570-150374570 on Build GRCh38
rs765856800 Chr.5: 150374582-150374582 on Build GRCh38
rs112039991 Chr.5: 150374653-150374653 on Build GRCh38
rs760851298 Chr.5: 150374749-150374749 on Build GRCh38
rs756453111 Chr.5: 150374603-150374603 on Build GRCh38
rs774997168 Chr.5: 150374703-150374703 on Build GRCh38
rs114326915 Chr.5: 150374762-150374762 on Build GRCh38
rs772235723 Chr.5: 150374673-150374673 on Build GRCh38
rs760076793 Chr.5: 150374776-150374776 on Build GRCh38
rs762624486 Chr.5: 150374695-150374695 on Build GRCh38
rs1009327429 Chr.5: 150374748-150374748 on Build GRCh38
rs544776908 Chr.5: 150374642-150374642 on Build GRCh38
rs748059590 Chr.5: 150374675-150374675 on Build GRCh38
rs760292610 Chr.5: 150374706-150374706 on Build GRCh38
rs771702524 Chr.5: 150374771-150374771 on Build GRCh38
rs761683030 Chr.5: 150374639-150374639 on Build GRCh38
rs746636582 Chr.5: 150374606-150374607 on Build GRCh38
rs373402474 Chr.5: 150374583-150374583 on Build GRCh38
rs772696466 Chr.5: 150374701-150374701 on Build GRCh38
rs774138891 Chr.5: 150374763-150374763 on Build GRCh38
rs367576572 Chr.5: 150374641-150374641 on Build GRCh38
rs772895657 Chr.5: 150374640-150374640 on Build GRCh38
rs997547513 Chr.5: 150374778-150374778 on Build GRCh38
rs201429329 Chr.5: 150374769-150374769 on Build GRCh38
rs753254761 Chr.5: 150374599-150374599 on Build GRCh38
rs374052117 Chr.5: 150374581-150374581 on Build GRCh38
rs778144940 Chr.5: 150374658-150374658 on Build GRCh38
rs759026634 Chr.5: 150374647-150374647 on Build GRCh38
rs757511240 Chr.5: 150374721-150374721 on Build GRCh38
rs774778131 Chr.5: 150374655-150374655 on Build GRCh38
rs751252576 Chr.5: 150374646-150374646 on Build GRCh38
rs773162787 Chr.5: 150374741-150374741 on Build GRCh38
rs764508712 Chr.5: 150374708-150374708 on Build GRCh38
rs745940724 Chr.5: 150374624-150374624 on Build GRCh38
rs756584594 Chr.5: 150374657-150374657 on Build GRCh38
rs553190321 Chr.5: 150374586-150374586 on Build GRCh38
rs770762508 Chr.5: 150374698-150374698 on Build GRCh38
rs754413205 Chr.5: 150374718-150374718 on Build GRCh38
rs769604188 Chr.5: 150374676-150374676 on Build GRCh38
rs560713844 Chr.5: 150374745-150374745 on Build GRCh38
rs373091130 Chr.5: 150374734-150374734 on Build GRCh38
rs771075643 Chr.5: 150374618-150374618 on Build GRCh38
rs778031599 Chr.5: 150374608-150374608 on Build GRCh38
rs777465512 Chr.5: 150374567-150374568 on Build GRCh38
rs150608282 Chr.5: 150374773-150374773 on Build GRCh38
rs370636217 Chr.5: 150374580-150374580 on Build GRCh38
rs75181211 Chr.5: 150374666-150374666 on Build GRCh38
rs149825269 Chr.5: 150374661-150374661 on Build GRCh38
rs749463438 Chr.5: 150374613-150374613 on Build GRCh38
rs749270349 Chr.5: 150374761-150374761 on Build GRCh38
rs566906192 Chr.5: 150374575-150374575 on Build GRCh38
rs780283141 Chr.5: 150374674-150374674 on Build GRCh38
rs376097692 Chr.5: 150374753-150374753 on Build GRCh38
rs766948726 Chr.5: 150374702-150374702 on Build GRCh38
rs776658410 Chr.5: 150374629-150374629 on Build GRCh38
rs776747398 Chr.5: 150374572-150374572 on Build GRCh38
rs554646790 Chr.5: 150374552-150374552 on Build GRCh38
rs759220649 Chr.5: 150374700-150374700 on Build GRCh38
rs151344568 Chr.5: 150374619-150374619 on Build GRCh38
rs189476787 Chr.5: 150374663-150374663 on Build GRCh38
rs760636303 Chr.5: 150374567-150374567 on Build GRCh38
rs151344569 Chr.5: 150374775-150374775 on Build GRCh38
rs780479645 Chr.5: 150374742-150374742 on Build GRCh38

View Full Product Details