Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73892025-73892282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF410
Entrez Gene ID
57862
Gene Name
zinc finger protein 410
Gene Aliases
APA-1, APA1
UniGene
Hs.270869
Gene Chromosome Location
Chr.14: 73886615-73932288 on Build GRCh38
Transcripts
NM_001242924.1 NM_001242926.1 NM_001242927.1 NM_001242928.1 NM_021188.2 NR_040251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815280 c.8C>T p.S3L Chr.14: 73892183-73892183 on Build GRCh38
6486508 c.24C>G p.S8S Chr.14: 73892199-73892199 on Build GRCh38
6345451 c.15G>T p.E5D Chr.14: 73892190-73892190 on Build GRCh38
5689400 c.31G>T p.E11* Chr.14: 73892206-73892206 on Build GRCh38
4430394 c.11A>G p.D4G Chr.14: 73892186-73892186 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764447000 Chr.14: 73892122-73892122 on Build GRCh38
rs756698259 Chr.14: 73892158-73892158 on Build GRCh38
rs375161976 Chr.14: 73892193-73892193 on Build GRCh38
rs368486670 Chr.14: 73892211-73892211 on Build GRCh38
rs778568081 Chr.14: 73892061-73892061 on Build GRCh38
rs757999945 Chr.14: 73892125-73892130 on Build GRCh38
rs745605563 Chr.14: 73892226-73892226 on Build GRCh38
rs77953765 Chr.14: 73892102-73892102 on Build GRCh38
rs779549092 Chr.14: 73892247-73892248 on Build GRCh38
rs746859689 Chr.14: 73892081-73892081 on Build GRCh38
rs772734308 Chr.14: 73892067-73892067 on Build GRCh38
rs999953038 Chr.14: 73892059-73892059 on Build GRCh38
rs762933624 Chr.14: 73892120-73892120 on Build GRCh38
rs776362558 Chr.14: 73892085-73892085 on Build GRCh38
rs746664764 Chr.14: 73892201-73892201 on Build GRCh38
rs752318148 Chr.14: 73892178-73892178 on Build GRCh38
rs780971389 Chr.14: 73892222-73892222 on Build GRCh38
rs1034558384 Chr.14: 73892110-73892110 on Build GRCh38
rs769653093 Chr.14: 73892092-73892092 on Build GRCh38
rs750747095 Chr.14: 73892151-73892151 on Build GRCh38
rs779651359 Chr.14: 73892161-73892161 on Build GRCh38
rs761264356 Chr.14: 73892097-73892097 on Build GRCh38
rs772634695 Chr.14: 73892064-73892064 on Build GRCh38
rs559660624 Chr.14: 73892091-73892091 on Build GRCh38
rs747549608 Chr.14: 73892062-73892062 on Build GRCh38
rs371715301 Chr.14: 73892142-73892142 on Build GRCh38
rs189104598 Chr.14: 73892082-73892082 on Build GRCh38
rs768598339 Chr.14: 73892246-73892246 on Build GRCh38
rs757814091 Chr.14: 73892190-73892190 on Build GRCh38
rs769785228 Chr.14: 73892232-73892232 on Build GRCh38
rs891662104 Chr.14: 73892066-73892066 on Build GRCh38
rs201800416 Chr.14: 73892247-73892247 on Build GRCh38
rs756646778 Chr.14: 73892057-73892057 on Build GRCh38
rs762289142 Chr.14: 73892144-73892144 on Build GRCh38
rs768022764 Chr.14: 73892147-73892147 on Build GRCh38
rs918746554 Chr.14: 73892235-73892235 on Build GRCh38
rs909332659 Chr.14: 73892076-73892076 on Build GRCh38
rs775288721 Chr.14: 73892241-73892241 on Build GRCh38

View Full Product Details