Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54228120-54228379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP3
Entrez Gene ID
23648
Gene Name
single stranded DNA binding protein 3
Gene Aliases
CSDP, SSDP, SSDP1
UniGene
Hs.476706
Gene Chromosome Location
Chr.1: 54225431-54413479 on Build GRCh38
Transcripts
NM_001009955.3 NM_018070.4 NM_145716.3 XM_006710545.3 XM_017000897.1 XM_017000898.1 XM_017000899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2193708 c.1041C>T p.S347S Chr.1: 54228270-54228270 on Build GRCh38
3764199 c.1033C>T p.L345F Chr.1: 54228278-54228278 on Build GRCh38
1584645 c.1040C>A p.S347Y Chr.1: 54228352-54228352 on Build GRCh38
2193706 c.1051G>A p.D351N Chr.1: 54228260-54228260 on Build GRCh38
3689766 c.1011C>T p.D337D Chr.1: 54228300-54228300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372030814 Chr.1: 54228246-54228246 on Build GRCh38
rs766705537 Chr.1: 54228260-54228260 on Build GRCh38
rs199631322 Chr.1: 54228256-54228256 on Build GRCh38
rs763247696 Chr.1: 54228230-54228230 on Build GRCh38
rs990834131 Chr.1: 54228191-54228191 on Build GRCh38
rs370231297 Chr.1: 54228218-54228218 on Build GRCh38
rs751443452 Chr.1: 54228153-54228153 on Build GRCh38
rs750935705 Chr.1: 54228248-54228248 on Build GRCh38
rs763937819 Chr.1: 54228145-54228145 on Build GRCh38
rs558695729 Chr.1: 54228144-54228144 on Build GRCh38
rs959423219 Chr.1: 54228188-54228188 on Build GRCh38
rs922373364 Chr.1: 54228236-54228236 on Build GRCh38
rs149091689 Chr.1: 54228270-54228270 on Build GRCh38
rs553646788 Chr.1: 54228299-54228299 on Build GRCh38
rs774470060 Chr.1: 54228242-54228242 on Build GRCh38
rs978404222 Chr.1: 54228202-54228202 on Build GRCh38
rs778884724 Chr.1: 54228275-54228277 on Build GRCh38
rs267598654 Chr.1: 54228271-54228271 on Build GRCh38
rs895652154 Chr.1: 54228175-54228175 on Build GRCh38
rs755795241 Chr.1: 54228298-54228298 on Build GRCh38
rs761905061 Chr.1: 54228245-54228245 on Build GRCh38
rs777431651 Chr.1: 54228276-54228276 on Build GRCh38
rs757269839 Chr.1: 54228189-54228189 on Build GRCh38
rs770366367 Chr.1: 54228219-54228219 on Build GRCh38
rs745807258 Chr.1: 54228341-54228341 on Build GRCh38
rs961270160 Chr.1: 54228165-54228165 on Build GRCh38
rs200431838 Chr.1: 54228240-54228240 on Build GRCh38
rs151171870 Chr.1: 54228300-54228300 on Build GRCh38
rs576666955 Chr.1: 54228351-54228351 on Build GRCh38
rs924178794 Chr.1: 54228209-54228209 on Build GRCh38
rs1035106501 Chr.1: 54228176-54228176 on Build GRCh38
rs780637364 Chr.1: 54228336-54228336 on Build GRCh38
rs775487322 Chr.1: 54228226-54228226 on Build GRCh38
rs139138239 Chr.1: 54228217-54228217 on Build GRCh38
rs568357592 Chr.1: 54228201-54228201 on Build GRCh38
rs145221324 Chr.1: 54228261-54228261 on Build GRCh38
rs114408769 Chr.1: 54228225-54228225 on Build GRCh38

View Full Product Details