Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53166245-53166470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF665
Entrez Gene ID
79788
Gene Name
zinc finger protein 665
Gene Aliases
ZFP160L
UniGene
Hs.745230
Gene Chromosome Location
Chr.19: 53163299-53193404 on Build GRCh38
Transcripts
NM_024733.3 XM_005259266.4 XM_011527325.2 XM_011527327.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
248618 c.207G>A p.A69A Chr.19: 53166283-53166283 on Build GRCh38
6832174 c.11C>T p.A4V Chr.19: 53166284-53166284 on Build GRCh38
7105553 c.193A>C p.N65H Chr.19: 53166297-53166297 on Build GRCh38
248617 c.12G>A p.A4A Chr.19: 53166283-53166283 on Build GRCh38
3672999 c.3G>T p.M1I Chr.19: 53166292-53166292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749992210 Chr.19: 53166395-53166395 on Build GRCh38
rs779863071 Chr.19: 53166322-53166322 on Build GRCh38
rs915123478 Chr.19: 53166418-53166418 on Build GRCh38
rs764720509 Chr.19: 53166299-53166299 on Build GRCh38
rs561977125 Chr.19: 53166338-53166338 on Build GRCh38
rs545877487 Chr.19: 53166275-53166275 on Build GRCh38
rs771182587 Chr.19: 53166328-53166328 on Build GRCh38
rs368697071 Chr.19: 53166319-53166319 on Build GRCh38
rs185879478 Chr.19: 53166283-53166283 on Build GRCh38
rs746957820 Chr.19: 53166321-53166330 on Build GRCh38
rs1004567662 Chr.19: 53166285-53166285 on Build GRCh38
rs745976247 Chr.19: 53166344-53166344 on Build GRCh38
rs777654533 Chr.19: 53166430-53166430 on Build GRCh38
rs1013514462 Chr.19: 53166312-53166312 on Build GRCh38
rs763838017 Chr.19: 53166284-53166284 on Build GRCh38
rs748623042 Chr.19: 53166313-53166313 on Build GRCh38
rs1015580344 Chr.19: 53166360-53166360 on Build GRCh38
rs12460233 Chr.19: 53166400-53166400 on Build GRCh38
rs757785534 Chr.19: 53166307-53166307 on Build GRCh38
rs543368305 Chr.19: 53166306-53166306 on Build GRCh38
rs781731850 Chr.19: 53166309-53166309 on Build GRCh38
rs374585821 Chr.19: 53166294-53166294 on Build GRCh38
rs368323920 Chr.19: 53166274-53166274 on Build GRCh38
rs767497325 Chr.19: 53166280-53166280 on Build GRCh38
rs775529570 Chr.19: 53166368-53166368 on Build GRCh38
rs371699472 Chr.19: 53166323-53166323 on Build GRCh38
rs771997272 Chr.19: 53166356-53166356 on Build GRCh38
rs533092142 Chr.19: 53166433-53166433 on Build GRCh38
rs528884984 Chr.19: 53166376-53166376 on Build GRCh38
rs985718086 Chr.19: 53166414-53166414 on Build GRCh38
rs756889208 Chr.19: 53166296-53166296 on Build GRCh38
rs191726004 Chr.19: 53166386-53166386 on Build GRCh38
rs376454561 Chr.19: 53166385-53166385 on Build GRCh38
rs758509426 Chr.19: 53166431-53166431 on Build GRCh38
rs751492177 Chr.19: 53166378-53166378 on Build GRCh38

View Full Product Details