Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19835530-19835763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYM5
Entrez Gene ID
9205
Gene Name
zinc finger MYM-type containing 5
Gene Aliases
HSPC050, MYM, ZNF198L1, ZNF237
UniGene
Hs.530988
Gene Chromosome Location
Chr.13: 19823482-19863636 on Build GRCh38
Transcripts
NM_001142684.1 XM_005266593.3 XM_005266594.2 XM_006719894.2 XM_011535309.1 XM_011535310.1 XM_017020849.1 XM_017020850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3399268 c.1093T>C p.C365R Chr.13: 19835635-19835635 on Build GRCh38
6827569 c.1150G>A p.G384R Chr.13: 19835578-19835578 on Build GRCh38
6355769 c.1086T>C p.S362S Chr.13: 19835642-19835642 on Build GRCh38
3467593 c.1163C>T p.P388L Chr.13: 19835565-19835565 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs921771175 Chr.13: 19835672-19835672 on Build GRCh38
rs766821743 Chr.13: 19835557-19835557 on Build GRCh38
rs780072268 Chr.13: 19835610-19835610 on Build GRCh38
rs747796183 Chr.13: 19835636-19835636 on Build GRCh38
rs912596520 Chr.13: 19835617-19835617 on Build GRCh38
rs372900063 Chr.13: 19835724-19835724 on Build GRCh38
rs369951273 Chr.13: 19835686-19835686 on Build GRCh38
rs539772012 Chr.13: 19835573-19835573 on Build GRCh38
rs773587673 Chr.13: 19835649-19835649 on Build GRCh38
rs753293798 Chr.13: 19835653-19835654 on Build GRCh38
rs368252781 Chr.13: 19835555-19835555 on Build GRCh38
rs534076583 Chr.13: 19835685-19835685 on Build GRCh38
rs758667322 Chr.13: 19835571-19835571 on Build GRCh38
rs756768755 Chr.13: 19835713-19835716 on Build GRCh38
rs993109242 Chr.13: 19835560-19835560 on Build GRCh38
rs750197198 Chr.13: 19835575-19835575 on Build GRCh38
rs749245671 Chr.13: 19835612-19835612 on Build GRCh38
rs752916575 Chr.13: 19835570-19835570 on Build GRCh38
rs765277333 Chr.13: 19835680-19835680 on Build GRCh38
rs773836607 Chr.13: 19835592-19835592 on Build GRCh38
rs200991329 Chr.13: 19835644-19835644 on Build GRCh38
rs756136097 Chr.13: 19835597-19835597 on Build GRCh38
rs566948933 Chr.13: 19835671-19835671 on Build GRCh38
rs776783051 Chr.13: 19835563-19835563 on Build GRCh38
rs189039493 Chr.13: 19835664-19835664 on Build GRCh38
rs759463821 Chr.13: 19835564-19835564 on Build GRCh38
rs200196724 Chr.13: 19835614-19835614 on Build GRCh38
rs754913572 Chr.13: 19835717-19835717 on Build GRCh38
rs747313731 Chr.13: 19835658-19835658 on Build GRCh38
rs765351405 Chr.13: 19835567-19835567 on Build GRCh38
rs753768724 Chr.13: 19835708-19835708 on Build GRCh38
rs759943322 Chr.13: 19835692-19835692 on Build GRCh38
rs923960123 Chr.13: 19835623-19835623 on Build GRCh38
rs1042434097 Chr.13: 19835619-19835619 on Build GRCh38
rs778463818 Chr.13: 19835633-19835633 on Build GRCh38
rs764046838 Chr.13: 19835646-19835647 on Build GRCh38
rs756042699 Chr.13: 19835702-19835702 on Build GRCh38
rs766185159 Chr.13: 19835705-19835705 on Build GRCh38
rs760645318 Chr.13: 19835560-19835562 on Build GRCh38

View Full Product Details