Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33453300-33453525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARS
Entrez Gene ID
6897
Gene Name
threonyl-tRNA synthetase
Gene Aliases
ThrRS
UniGene
Hs.481860
Gene Chromosome Location
Chr.5: 33440696-33468091 on Build GRCh38
Transcripts
NM_001258437.1 NM_001258438.1 NM_152295.4 NR_047676.1 NR_047677.1 NR_047678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6300534 c.442G>T p.E148* Chr.5: 33453401-33453401 on Build GRCh38
1067342 c.383A>G p.D128G Chr.5: 33453342-33453342 on Build GRCh38
5996124 c.370A>G p.N124D Chr.5: 33453329-33453329 on Build GRCh38
5344927 c.371A>T p.N124I Chr.5: 33453330-33453330 on Build GRCh38
185937 c.392G>A p.R131H Chr.5: 33453351-33453351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767290886 Chr.5: 33453351-33453351 on Build GRCh38
rs774448715 Chr.5: 33453450-33453450 on Build GRCh38
rs753448647 Chr.5: 33453371-33453371 on Build GRCh38
rs759139113 Chr.5: 33453402-33453402 on Build GRCh38
rs202220873 Chr.5: 33453436-33453436 on Build GRCh38
rs936488118 Chr.5: 33453497-33453497 on Build GRCh38
rs755358781 Chr.5: 33453410-33453410 on Build GRCh38
rs759432043 Chr.5: 33453340-33453340 on Build GRCh38
rs756701557 Chr.5: 33453391-33453391 on Build GRCh38
rs771876897 Chr.5: 33453461-33453461 on Build GRCh38
rs763653176 Chr.5: 33453366-33453366 on Build GRCh38
rs371779976 Chr.5: 33453425-33453425 on Build GRCh38
rs774360891 Chr.5: 33453336-33453336 on Build GRCh38
rs890803196 Chr.5: 33453493-33453493 on Build GRCh38
rs375341932 Chr.5: 33453438-33453438 on Build GRCh38
rs760419283 Chr.5: 33453354-33453354 on Build GRCh38
rs745846263 Chr.5: 33453451-33453451 on Build GRCh38
rs752407851 Chr.5: 33453353-33453353 on Build GRCh38
rs780546165 Chr.5: 33453401-33453401 on Build GRCh38
rs777799549 Chr.5: 33453429-33453429 on Build GRCh38
rs749380627 Chr.5: 33453430-33453430 on Build GRCh38
rs538061732 Chr.5: 33453490-33453490 on Build GRCh38
rs569001039 Chr.5: 33453395-33453395 on Build GRCh38

View Full Product Details