Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66994055-66994294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBDS
Entrez Gene ID
51119
Gene Name
SBDS ribosome assembly guanine nucleotide exchange factor
Gene Aliases
CGI-97, SDS, SWDS
UniGene
Hs.110445
Gene Chromosome Location
Chr.7: 66987703-66995601 on Build GRCh38
Transcripts
NM_016038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3995668 c.257A>G p.Q86R Chr.7: 66994213-66994213 on Build GRCh38
1313244 c.213C>T p.L71L Chr.7: 66994257-66994257 on Build GRCh38
3762810 c.201A>G p.K67K Chr.7: 66994269-66994269 on Build GRCh38
3778541 c.205G>A p.E69K Chr.7: 66994265-66994265 on Build GRCh38
5457346 c.192G>T p.Q64H Chr.7: 66994278-66994278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961823323 Chr.7: 66994176-66994176 on Build GRCh38
rs181692878 Chr.7: 66994153-66994153 on Build GRCh38
rs574544072 Chr.7: 66994185-66994185 on Build GRCh38
rs374263583 Chr.7: 66994165-66994165 on Build GRCh38
rs772119912 Chr.7: 66994228-66994228 on Build GRCh38
rs752076745 Chr.7: 66994169-66994169 on Build GRCh38
rs953503293 Chr.7: 66994208-66994208 on Build GRCh38
rs571241815 Chr.7: 66994087-66994087 on Build GRCh38
rs765553121 Chr.7: 66994163-66994163 on Build GRCh38
rs1061695 Chr.7: 66994269-66994269 on Build GRCh38
rs537167027 Chr.7: 66994088-66994088 on Build GRCh38
rs760622455 Chr.7: 66994233-66994233 on Build GRCh38
rs1011914990 Chr.7: 66994124-66994124 on Build GRCh38
rs11763269 Chr.7: 66994158-66994158 on Build GRCh38
rs755088500 Chr.7: 66994152-66994152 on Build GRCh38
rs536015496 Chr.7: 66994164-66994164 on Build GRCh38
rs564159611 Chr.7: 66994252-66994252 on Build GRCh38
rs113993993 Chr.7: 66994210-66994210 on Build GRCh38
rs769235557 Chr.7: 66994249-66994249 on Build GRCh38
rs749210195 Chr.7: 66994184-66994184 on Build GRCh38
rs1044768408 Chr.7: 66994139-66994145 on Build GRCh38
rs775652243 Chr.7: 66994230-66994230 on Build GRCh38
rs4307226 Chr.7: 66994128-66994128 on Build GRCh38
rs746159940 Chr.7: 66994219-66994219 on Build GRCh38
rs372215536 Chr.7: 66994182-66994182 on Build GRCh38
rs780817749 Chr.7: 66994166-66994166 on Build GRCh38
rs368647191 Chr.7: 66994175-66994175 on Build GRCh38
rs186000847 Chr.7: 66994193-66994193 on Build GRCh38
rs1030333829 Chr.7: 66994194-66994194 on Build GRCh38
rs113993992 Chr.7: 66994211-66994211 on Build GRCh38
rs765645155 Chr.7: 66994266-66994266 on Build GRCh38

View Full Product Details