Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39150231-39150502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC6B
Entrez Gene ID
122553
Gene Name
trafficking protein particle complex 6B
Gene Aliases
TPC6
UniGene
Hs.13303
Gene Chromosome Location
Chr.14: 39147811-39170430 on Build GRCh38
Transcripts
NM_001079537.1 NM_177452.3 XM_011536409.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4385897 c.446-2_446-1insT p.? Chr.14: 39150382-39150383 on Build GRCh38
4438095 c.446-3_446-2insT p.? Chr.14: 39150383-39150384 on Build GRCh38
1580341 c.446-3delT p.? Chr.14: 39150384-39150384 on Build GRCh38
2196812 c.476A>T p.*159L Chr.14: 39150351-39150351 on Build GRCh38
5513035 c.446-4_446-3delTT p.? Chr.14: 39150384-39150385 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756513266 Chr.14: 39150384-39150384 on Build GRCh38
rs192358838 Chr.14: 39150406-39150406 on Build GRCh38
rs908663554 Chr.14: 39150460-39150460 on Build GRCh38
rs920059973 Chr.14: 39150412-39150412 on Build GRCh38
rs992936056 Chr.14: 39150336-39150336 on Build GRCh38
rs774733485 Chr.14: 39150348-39150348 on Build GRCh38
rs751259011 Chr.14: 39150310-39150311 on Build GRCh38
rs756635177 Chr.14: 39150384-39150385 on Build GRCh38
rs768067381 Chr.14: 39150367-39150367 on Build GRCh38
rs753241814 Chr.14: 39150404-39150404 on Build GRCh38
rs769919403 Chr.14: 39150320-39150320 on Build GRCh38
rs150886646 Chr.14: 39150384-39150384 on Build GRCh38
rs763694776 Chr.14: 39150395-39150395 on Build GRCh38
rs376059015 Chr.14: 39150342-39150342 on Build GRCh38
rs772178298 Chr.14: 39150313-39150313 on Build GRCh38
rs757020893 Chr.14: 39150422-39150422 on Build GRCh38
rs753264525 Chr.14: 39150374-39150374 on Build GRCh38
rs1003834034 Chr.14: 39150297-39150297 on Build GRCh38
rs895558467 Chr.14: 39150370-39150370 on Build GRCh38
rs74683746 Chr.14: 39150353-39150353 on Build GRCh38
rs982864234 Chr.14: 39150456-39150456 on Build GRCh38
rs753148660 Chr.14: 39150383-39150384 on Build GRCh38
rs188974892 Chr.14: 39150360-39150360 on Build GRCh38
rs775565275 Chr.14: 39150314-39150314 on Build GRCh38
rs773126055 Chr.14: 39150325-39150325 on Build GRCh38
rs931496584 Chr.14: 39150419-39150419 on Build GRCh38
rs981460881 Chr.14: 39150443-39150443 on Build GRCh38
rs549552968 Chr.14: 39150467-39150467 on Build GRCh38
rs753523357 Chr.14: 39150259-39150259 on Build GRCh38
rs1022636995 Chr.14: 39150349-39150349 on Build GRCh38
rs1032577573 Chr.14: 39150451-39150451 on Build GRCh38
rs1013711875 Chr.14: 39150378-39150378 on Build GRCh38
rs536392862 Chr.14: 39150446-39150446 on Build GRCh38
rs888148461 Chr.14: 39150296-39150296 on Build GRCh38
rs754431393 Chr.14: 39150316-39150316 on Build GRCh38
rs372801793 Chr.14: 39150317-39150317 on Build GRCh38

View Full Product Details