Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3615112-3615373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHPK
Entrez Gene ID
23729
Gene Name
sedoheptulokinase
Gene Aliases
CARKL, SHK
UniGene
Hs.579217
Gene Chromosome Location
Chr.17: 3608262-3636322 on Build GRCh38
Transcripts
NM_013276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6289575 c.1011G>A p.W337* Chr.17: 3615350-3615350 on Build GRCh38
6777966 c.1000C>A p.L334M Chr.17: 3615361-3615361 on Build GRCh38
6455919 c.990C>T p.F330F Chr.17: 3615371-3615371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918535313 Chr.17: 3615177-3615177 on Build GRCh38
rs895354710 Chr.17: 3615222-3615222 on Build GRCh38
rs375193270 Chr.17: 3615257-3615257 on Build GRCh38
rs182539973 Chr.17: 3615236-3615236 on Build GRCh38
rs931229186 Chr.17: 3615183-3615183 on Build GRCh38
rs1055211910 Chr.17: 3615171-3615171 on Build GRCh38
rs546941167 Chr.17: 3615242-3615242 on Build GRCh38
rs1031063733 Chr.17: 3615251-3615251 on Build GRCh38
rs996297737 Chr.17: 3615282-3615282 on Build GRCh38
rs754457483 Chr.17: 3615297-3615297 on Build GRCh38
rs186675577 Chr.17: 3615136-3615136 on Build GRCh38
rs115347888 Chr.17: 3615152-3615152 on Build GRCh38
rs1016298265 Chr.17: 3615252-3615252 on Build GRCh38
rs538801716 Chr.17: 3615250-3615250 on Build GRCh38
rs368391366 Chr.17: 3615312-3615312 on Build GRCh38
rs769640623 Chr.17: 3615325-3615325 on Build GRCh38
rs899236552 Chr.17: 3615266-3615266 on Build GRCh38
rs925655020 Chr.17: 3615328-3615328 on Build GRCh38
rs773119171 Chr.17: 3615346-3615346 on Build GRCh38
rs567162129 Chr.17: 3615243-3615243 on Build GRCh38
rs551607681 Chr.17: 3615154-3615154 on Build GRCh38
rs190322162 Chr.17: 3615218-3615218 on Build GRCh38
rs144011808 Chr.17: 3615287-3615287 on Build GRCh38
rs571516429 Chr.17: 3615156-3615156 on Build GRCh38
rs778865879 Chr.17: 3615302-3615302 on Build GRCh38
rs773202876 Chr.17: 3615235-3615235 on Build GRCh38
rs537928040 Chr.17: 3615303-3615303 on Build GRCh38
rs954573629 Chr.17: 3615288-3615288 on Build GRCh38
rs567128152 Chr.17: 3615230-3615230 on Build GRCh38
rs370963928 Chr.17: 3615315-3615315 on Build GRCh38
rs752984096 Chr.17: 3615286-3615286 on Build GRCh38
rs765882675 Chr.17: 3615345-3615345 on Build GRCh38
rs780370209 Chr.17: 3615316-3615316 on Build GRCh38
rs978382468 Chr.17: 3615326-3615326 on Build GRCh38
rs372221691 Chr.17: 3615162-3615162 on Build GRCh38
rs757359189 Chr.17: 3615301-3615301 on Build GRCh38
rs937088195 Chr.17: 3615345-3615345 on Build GRCh38
rs374663056 Chr.17: 3615319-3615319 on Build GRCh38

View Full Product Details